Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43194292-43194565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNDBP1
Entrez Gene ID
23582
Gene Name
cyclin D1 binding protein 1
Gene Aliases
DIP1, GCIP, HHM
UniGene
Hs.36794
Gene Chromosome Location
Chr.15: 43185118-43197177 on Build GRCh38
Transcripts
NM_012142.4 NR_027513.2 NR_027514.2 NR_045998.1 NR_045999.1 XM_006720448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1741371 c.958G>C p.E320Q Chr.15: 43194451-43194451 on Build GRCh38
212974 c.925G>C p.A309P Chr.15: 43194418-43194418 on Build GRCh38
3386876 c.951G>A p.K317K Chr.15: 43194444-43194444 on Build GRCh38
4573038 c.944T>C p.L315S Chr.15: 43194437-43194437 on Build GRCh38
1478131 c.958G>A p.E320K Chr.15: 43194451-43194451 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368655210 Chr.15: 43194379-43194379 on Build GRCh38
rs371988422 Chr.15: 43194395-43194395 on Build GRCh38
rs188597001 Chr.15: 43194503-43194503 on Build GRCh38
rs377272929 Chr.15: 43194458-43194458 on Build GRCh38
rs767698971 Chr.15: 43194442-43194442 on Build GRCh38
rs138245123 Chr.15: 43194433-43194433 on Build GRCh38
rs549998225 Chr.15: 43194476-43194476 on Build GRCh38
rs148515458 Chr.15: 43194521-43194521 on Build GRCh38
rs900569137 Chr.15: 43194429-43194429 on Build GRCh38
rs757025058 Chr.15: 43194459-43194459 on Build GRCh38
rs758394504 Chr.15: 43194474-43194474 on Build GRCh38
rs879662236 Chr.15: 43194317-43194317 on Build GRCh38
rs78728860 Chr.15: 43194420-43194420 on Build GRCh38
rs769207488 Chr.15: 43194383-43194383 on Build GRCh38
rs995039022 Chr.15: 43194445-43194445 on Build GRCh38
rs115573144 Chr.15: 43194419-43194419 on Build GRCh38
rs768415359 Chr.15: 43194365-43194365 on Build GRCh38
rs752914840 Chr.15: 43194448-43194448 on Build GRCh38
rs1023340004 Chr.15: 43194391-43194391 on Build GRCh38
rs374575032 Chr.15: 43194370-43194370 on Build GRCh38
rs746765400 Chr.15: 43194498-43194498 on Build GRCh38
rs971328222 Chr.15: 43194399-43194400 on Build GRCh38
rs369982564 Chr.15: 43194495-43194495 on Build GRCh38
rs763376168 Chr.15: 43194417-43194417 on Build GRCh38
rs1003145014 Chr.15: 43194416-43194416 on Build GRCh38
rs753533080 Chr.15: 43194383-43194384 on Build GRCh38
rs1008849069 Chr.15: 43194359-43194359 on Build GRCh38
rs560808540 Chr.15: 43194504-43194504 on Build GRCh38
rs890418830 Chr.15: 43194332-43194332 on Build GRCh38
rs531387107 Chr.15: 43194460-43194460 on Build GRCh38
rs952182859 Chr.15: 43194520-43194520 on Build GRCh38
rs575106343 Chr.15: 43194473-43194473 on Build GRCh38
rs375689084 Chr.15: 43194402-43194403 on Build GRCh38

View Full Product Details