Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152652679-152652904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRQ
Entrez Gene ID
55879
Gene Name
gamma-aminobutyric acid type A receptor theta subunit
Gene Aliases
THETA
UniGene
Hs.283081
Gene Chromosome Location
Chr.X: 152637895-152659827 on Build GRCh38
Transcripts
NM_018558.3 XM_011531184.2

SNP Detail

SNP ID Chromosome Location
rs372512790 Chr.X: 152652735-152652735 on Build GRCh38
rs782272412 Chr.X: 152652716-152652716 on Build GRCh38
rs372765591 Chr.X: 152652864-152652864 on Build GRCh38
rs782106882 Chr.X: 152652879-152652879 on Build GRCh38
rs142223987 Chr.X: 152652836-152652836 on Build GRCh38
rs143687531 Chr.X: 152652701-152652701 on Build GRCh38
rs138966264 Chr.X: 152652781-152652781 on Build GRCh38
rs781853975 Chr.X: 152652825-152652825 on Build GRCh38
rs782057430 Chr.X: 152652851-152652851 on Build GRCh38
rs77796656 Chr.X: 152652757-152652757 on Build GRCh38
rs80113450 Chr.X: 152652840-152652840 on Build GRCh38
rs781991760 Chr.X: 152652807-152652807 on Build GRCh38
rs782426371 Chr.X: 152652802-152652802 on Build GRCh38
rs782603061 Chr.X: 152652711-152652711 on Build GRCh38
rs781947829 Chr.X: 152652850-152652850 on Build GRCh38
rs782396610 Chr.X: 152652854-152652854 on Build GRCh38
rs961027252 Chr.X: 152652856-152652856 on Build GRCh38
rs782630695 Chr.X: 152652725-152652725 on Build GRCh38
rs368659256 Chr.X: 152652830-152652830 on Build GRCh38
rs889394845 Chr.X: 152652706-152652706 on Build GRCh38
rs781895777 Chr.X: 152652812-152652812 on Build GRCh38
rs782416882 Chr.X: 152652743-152652743 on Build GRCh38
rs782397568 Chr.X: 152652839-152652839 on Build GRCh38
rs782197983 Chr.X: 152652831-152652831 on Build GRCh38
rs782742963 Chr.X: 152652881-152652881 on Build GRCh38
rs782513983 Chr.X: 152652722-152652722 on Build GRCh38
rs782766146 Chr.X: 152652823-152652823 on Build GRCh38
rs141027732 Chr.X: 152652712-152652712 on Build GRCh38
rs199649217 Chr.X: 152652764-152652764 on Build GRCh38
rs78484935 Chr.X: 152652744-152652744 on Build GRCh38
rs3810651 Chr.X: 152652814-152652814 on Build GRCh38
rs782755581 Chr.X: 152652843-152652844 on Build GRCh38
rs77878508 Chr.X: 152652719-152652719 on Build GRCh38
rs782768514 Chr.X: 152652765-152652765 on Build GRCh38
rs781958098 Chr.X: 152652706-152652707 on Build GRCh38
rs782106906 Chr.X: 152652815-152652815 on Build GRCh38
rs377031308 Chr.X: 152652739-152652739 on Build GRCh38
rs200635320 Chr.X: 152652703-152652703 on Build GRCh38
rs188183099 Chr.X: 152652822-152652822 on Build GRCh38
rs782073292 Chr.X: 152652736-152652736 on Build GRCh38
rs5925198 Chr.X: 152652846-152652846 on Build GRCh38
rs782431029 Chr.X: 152652835-152652835 on Build GRCh38
rs782349764 Chr.X: 152652849-152652849 on Build GRCh38
rs782062554 Chr.X: 152652829-152652829 on Build GRCh38

View Full Product Details