Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 20846004-20846277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKAL1
Entrez Gene ID
54901
Gene Name
CDK5 regulatory subunit associated protein 1 like 1
Gene Aliases
-
UniGene
Hs.657604
Gene Chromosome Location
Chr.6: 20534457-21232404 on Build GRCh38
Transcripts
NM_017774.3 XM_011514718.1 XM_011514719.2 XM_017010986.1 XM_017010987.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3928248 c.692C>T p.A231V Chr.6: 20846128-20846128 on Build GRCh38
1742666 c.739C>T p.Q247* Chr.6: 20846175-20846175 on Build GRCh38
5756816 c.701C>T p.P234L Chr.6: 20846137-20846137 on Build GRCh38
4791475 c.670A>G p.K224E Chr.6: 20846106-20846106 on Build GRCh38
5869854 c.701C>A p.P234Q Chr.6: 20846137-20846137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751393112 Chr.6: 20846043-20846043 on Build GRCh38
rs762575761 Chr.6: 20846165-20846165 on Build GRCh38
rs767380491 Chr.6: 20846215-20846215 on Build GRCh38
rs775048563 Chr.6: 20846148-20846148 on Build GRCh38
rs772381613 Chr.6: 20846114-20846114 on Build GRCh38
rs762612512 Chr.6: 20846046-20846046 on Build GRCh38
rs1005914526 Chr.6: 20846228-20846228 on Build GRCh38
rs768321729 Chr.6: 20846170-20846170 on Build GRCh38
rs201700133 Chr.6: 20846060-20846060 on Build GRCh38
rs747453960 Chr.6: 20846127-20846127 on Build GRCh38
rs769197136 Chr.6: 20846140-20846140 on Build GRCh38
rs761722084 Chr.6: 20846199-20846199 on Build GRCh38
rs779292741 Chr.6: 20846087-20846087 on Build GRCh38
rs780114614 Chr.6: 20846065-20846065 on Build GRCh38
rs755225142 Chr.6: 20846085-20846085 on Build GRCh38
rs146424266 Chr.6: 20846111-20846111 on Build GRCh38
rs767537890 Chr.6: 20846047-20846047 on Build GRCh38
rs761730957 Chr.6: 20846189-20846189 on Build GRCh38
rs868012877 Chr.6: 20846145-20846145 on Build GRCh38
rs570778266 Chr.6: 20846050-20846050 on Build GRCh38
rs780144368 Chr.6: 20846091-20846091 on Build GRCh38
rs112984088 Chr.6: 20846097-20846097 on Build GRCh38
rs774335981 Chr.6: 20846194-20846194 on Build GRCh38
rs143969595 Chr.6: 20846083-20846083 on Build GRCh38
rs773743090 Chr.6: 20846121-20846121 on Build GRCh38

View Full Product Details