Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30654659-30654904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHR2
Entrez Gene ID
1395
Gene Name
corticotropin releasing hormone receptor 2
Gene Aliases
CRF-RB, CRF2, CRFR2, HM-CRF
UniGene
Hs.546246
Gene Chromosome Location
Chr.7: 30651844-30700103 on Build GRCh38
Transcripts
NM_001202475.1 NM_001202481.1 NM_001202482.1 NM_001202483.1 NM_001883.4 XM_011515127.2 XM_011515128.2 XM_011515129.2 XM_017011751.1 XM_017011752.1

SNP Detail

SNP ID Chromosome Location
rs770188242 Chr.7: 30654806-30654807 on Build GRCh38
rs1017332892 Chr.7: 30654881-30654881 on Build GRCh38
rs138102503 Chr.7: 30654865-30654865 on Build GRCh38
rs761527526 Chr.7: 30654790-30654790 on Build GRCh38
rs760650154 Chr.7: 30654832-30654832 on Build GRCh38
rs752088562 Chr.7: 30654678-30654678 on Build GRCh38
rs1036082021 Chr.7: 30654826-30654826 on Build GRCh38
rs73085814 Chr.7: 30654806-30654806 on Build GRCh38
rs574360909 Chr.7: 30654707-30654707 on Build GRCh38
rs991609899 Chr.7: 30654734-30654734 on Build GRCh38
rs920337424 Chr.7: 30654828-30654828 on Build GRCh38
rs909052113 Chr.7: 30654690-30654690 on Build GRCh38
rs568709511 Chr.7: 30654781-30654781 on Build GRCh38
rs767278994 Chr.7: 30654799-30654799 on Build GRCh38
rs991896939 Chr.7: 30654709-30654709 on Build GRCh38
rs981981967 Chr.7: 30654776-30654776 on Build GRCh38
rs969698188 Chr.7: 30654774-30654774 on Build GRCh38
rs774508299 Chr.7: 30654771-30654771 on Build GRCh38
rs762463251 Chr.7: 30654882-30654884 on Build GRCh38
rs985008711 Chr.7: 30654681-30654681 on Build GRCh38
rs916525972 Chr.7: 30654710-30654710 on Build GRCh38
rs763203256 Chr.7: 30654691-30654691 on Build GRCh38
rs915894107 Chr.7: 30654763-30654763 on Build GRCh38

View Full Product Details