Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38222184-38222455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPF1
Entrez Gene ID
8193
Gene Name
double PHD fingers 1
Gene Aliases
BAF45b, NEUD4, neuro-d4
UniGene
Hs.631576
Gene Chromosome Location
Chr.19: 38211006-38229714 on Build GRCh38
Transcripts
NM_001135155.2 NM_001135156.2 NM_001289978.1 NM_004647.3 XM_005259289.2 XM_005259290.3 XM_005259292.4 XM_006723407.2 XM_006723408.3 XM_006723409.2 XM_011527356.1 XM_011527357.2 XM_011527358.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3201913 c.254A>G p.N85S Chr.19: 38222401-38222401 on Build GRCh38
3201915 c.227G>A p.R76H Chr.19: 38222428-38222428 on Build GRCh38
1204472 c.329G>T p.R110I Chr.19: 38222407-38222407 on Build GRCh38
4541694 c.223G>C p.A75P Chr.19: 38222432-38222432 on Build GRCh38
6332730 c.261G>A p.L87L Chr.19: 38222394-38222394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764120701 Chr.19: 38222410-38222410 on Build GRCh38
rs920542287 Chr.19: 38222374-38222374 on Build GRCh38
rs997121052 Chr.19: 38222263-38222263 on Build GRCh38
rs779780162 Chr.19: 38222319-38222319 on Build GRCh38
rs747785017 Chr.19: 38222346-38222346 on Build GRCh38
rs533919458 Chr.19: 38222222-38222222 on Build GRCh38
rs572481027 Chr.19: 38222209-38222209 on Build GRCh38
rs774580084 Chr.19: 38222418-38222418 on Build GRCh38
rs373603463 Chr.19: 38222364-38222364 on Build GRCh38
rs1037554838 Chr.19: 38222358-38222358 on Build GRCh38
rs367709872 Chr.19: 38222433-38222433 on Build GRCh38
rs778735938 Chr.19: 38222344-38222344 on Build GRCh38
rs952691972 Chr.19: 38222280-38222280 on Build GRCh38
rs140246182 Chr.19: 38222424-38222424 on Build GRCh38
rs375413164 Chr.19: 38222293-38222293 on Build GRCh38
rs757035609 Chr.19: 38222307-38222307 on Build GRCh38
rs200845328 Chr.19: 38222401-38222401 on Build GRCh38
rs748798814 Chr.19: 38222322-38222322 on Build GRCh38
rs961468590 Chr.19: 38222336-38222336 on Build GRCh38
rs768210796 Chr.19: 38222328-38222328 on Build GRCh38
rs544441230 Chr.19: 38222296-38222296 on Build GRCh38
rs143882522 Chr.19: 38222394-38222394 on Build GRCh38
rs12982549 Chr.19: 38222279-38222279 on Build GRCh38
rs763026633 Chr.19: 38222407-38222407 on Build GRCh38
rs780993479 Chr.19: 38222311-38222311 on Build GRCh38
rs756050206 Chr.19: 38222318-38222318 on Build GRCh38
rs771247053 Chr.19: 38222367-38222367 on Build GRCh38
rs777020703 Chr.19: 38222375-38222375 on Build GRCh38
rs1055593305 Chr.19: 38222411-38222411 on Build GRCh38
rs1041200245 Chr.19: 38222228-38222228 on Build GRCh38
rs371061615 Chr.19: 38222345-38222345 on Build GRCh38
rs1005550451 Chr.19: 38222286-38222286 on Build GRCh38
rs750379824 Chr.19: 38222316-38222316 on Build GRCh38
rs746427575 Chr.19: 38222387-38222387 on Build GRCh38
rs999458162 Chr.19: 38222223-38222223 on Build GRCh38
rs555170168 Chr.19: 38222262-38222262 on Build GRCh38

View Full Product Details