Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 120951120-120951389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLI2
Entrez Gene ID
2736
Gene Name
GLI family zinc finger 2
Gene Aliases
CJS, HPE9, PHS2, THP1, THP2
UniGene
Hs.111867
Gene Chromosome Location
Chr.2: 120797291-120992653 on Build GRCh38
Transcripts
NM_005270.4 XM_006712422.2 XM_011510969.2 XM_011510970.2 XM_011510971.2 XM_011510972.2 XM_011510973.2 XM_011510974.2 XM_017003818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
716023 c.342C>T p.P114P Chr.2: 120951330-120951330 on Build GRCh38
311477 c.305G>C p.R102P Chr.2: 120951293-120951293 on Build GRCh38
6087257 c.257C>A p.P86H Chr.2: 120951245-120951245 on Build GRCh38
7169909 c.378delG p.E127fs*15 Chr.2: 120951366-120951366 on Build GRCh38
3041548 c.289G>A p.D97N Chr.2: 120951277-120951277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050289427 Chr.2: 120951280-120951280 on Build GRCh38
rs771817329 Chr.2: 120951358-120951358 on Build GRCh38
rs960826347 Chr.2: 120951154-120951154 on Build GRCh38
rs940582075 Chr.2: 120951340-120951340 on Build GRCh38
rs775273286 Chr.2: 120951364-120951364 on Build GRCh38
rs767309313 Chr.2: 120951307-120951307 on Build GRCh38
rs767375449 Chr.2: 120951222-120951222 on Build GRCh38
rs749323334 Chr.2: 120951190-120951190 on Build GRCh38
rs755546542 Chr.2: 120951345-120951345 on Build GRCh38
rs748827715 Chr.2: 120951351-120951351 on Build GRCh38
rs374562179 Chr.2: 120951308-120951308 on Build GRCh38
rs753931354 Chr.2: 120951317-120951317 on Build GRCh38
rs780467517 Chr.2: 120951273-120951273 on Build GRCh38
rs556523182 Chr.2: 120951147-120951147 on Build GRCh38
rs764133019 Chr.2: 120951227-120951227 on Build GRCh38
rs773048090 Chr.2: 120951194-120951194 on Build GRCh38
rs895231080 Chr.2: 120951370-120951370 on Build GRCh38
rs745508607 Chr.2: 120951354-120951354 on Build GRCh38
rs532928261 Chr.2: 120951224-120951224 on Build GRCh38
rs778467271 Chr.2: 120951352-120951352 on Build GRCh38
rs905463731 Chr.2: 120951347-120951347 on Build GRCh38
rs779141145 Chr.2: 120951246-120951246 on Build GRCh38
rs988597060 Chr.2: 120951187-120951187 on Build GRCh38
rs370693310 Chr.2: 120951302-120951302 on Build GRCh38
rs540240518 Chr.2: 120951259-120951259 on Build GRCh38
rs574293436 Chr.2: 120951310-120951310 on Build GRCh38
rs780453822 Chr.2: 120951343-120951343 on Build GRCh38
rs886357294 Chr.2: 120951205-120951205 on Build GRCh38
rs140711756 Chr.2: 120951336-120951336 on Build GRCh38
rs368546026 Chr.2: 120951369-120951369 on Build GRCh38
rs775984106 Chr.2: 120951337-120951337 on Build GRCh38
rs536400122 Chr.2: 120951156-120951156 on Build GRCh38
rs368449972 Chr.2: 120951292-120951292 on Build GRCh38
rs151057041 Chr.2: 120951211-120951211 on Build GRCh38
rs759277639 Chr.2: 120951306-120951306 on Build GRCh38
rs370629096 Chr.2: 120951258-120951258 on Build GRCh38
rs148442092 Chr.2: 120951293-120951293 on Build GRCh38
rs568965080 Chr.2: 120951146-120951146 on Build GRCh38
rs143139718 Chr.2: 120951316-120951316 on Build GRCh38
rs779428379 Chr.2: 120951251-120951251 on Build GRCh38
rs748739602 Chr.2: 120951296-120951296 on Build GRCh38
rs781044065 Chr.2: 120951207-120951209 on Build GRCh38
rs913979797 Chr.2: 120951216-120951216 on Build GRCh38
rs748092148 Chr.2: 120951212-120951214 on Build GRCh38
rs758739873 Chr.2: 120951341-120951341 on Build GRCh38
rs866998741 Chr.2: 120951231-120951231 on Build GRCh38
rs139473153 Chr.2: 120951303-120951303 on Build GRCh38
rs199499256 Chr.2: 120951226-120951226 on Build GRCh38
rs578011257 Chr.2: 120951309-120951309 on Build GRCh38
rs199671413 Chr.2: 120951279-120951279 on Build GRCh38
rs762680489 Chr.2: 120951202-120951202 on Build GRCh38
rs757479194 Chr.2: 120951245-120951245 on Build GRCh38
rs781740772 Chr.2: 120951348-120951348 on Build GRCh38
rs750629679 Chr.2: 120951338-120951338 on Build GRCh38
rs374679473 Chr.2: 120951221-120951221 on Build GRCh38
rs766342321 Chr.2: 120951204-120951204 on Build GRCh38
rs754130740 Chr.2: 120951230-120951230 on Build GRCh38
rs769510029 Chr.2: 120951229-120951230 on Build GRCh38

View Full Product Details