Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174262711-174262954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO8
Entrez Gene ID
26269
Gene Name
F-box protein 8
Gene Aliases
DC10, FBS, FBX8
UniGene
Hs.76917
Gene Chromosome Location
Chr.4: 174236658-174284251 on Build GRCh38
Transcripts
NM_012180.2 XM_017008020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5017727 c.293G>A p.W98* Chr.4: 174262800-174262800 on Build GRCh38
4123476 c.317T>G p.L106R Chr.4: 174262776-174262776 on Build GRCh38
4684053 c.158T>C p.I53T Chr.4: 174262935-174262935 on Build GRCh38
289831 c.306G>A p.A102A Chr.4: 174262787-174262787 on Build GRCh38
208209 c.204C>T p.F68F Chr.4: 174262889-174262889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771489632 Chr.4: 174262788-174262788 on Build GRCh38
rs149560688 Chr.4: 174262817-174262817 on Build GRCh38
rs906977178 Chr.4: 174262811-174262811 on Build GRCh38
rs915602618 Chr.4: 174262761-174262761 on Build GRCh38
rs757032130 Chr.4: 174262868-174262868 on Build GRCh38
rs140770819 Chr.4: 174262928-174262928 on Build GRCh38
rs1011942594 Chr.4: 174262913-174262913 on Build GRCh38
rs565911806 Chr.4: 174262874-174262874 on Build GRCh38
rs760065820 Chr.4: 174262792-174262792 on Build GRCh38
rs545288874 Chr.4: 174262894-174262894 on Build GRCh38
rs760079454 Chr.4: 174262808-174262808 on Build GRCh38
rs769805068 Chr.4: 174262908-174262908 on Build GRCh38
rs763345340 Chr.4: 174262787-174262787 on Build GRCh38
rs778056634 Chr.4: 174262918-174262918 on Build GRCh38
rs763766386 Chr.4: 174262845-174262845 on Build GRCh38
rs772602892 Chr.4: 174262907-174262907 on Build GRCh38
rs147248654 Chr.4: 174262876-174262876 on Build GRCh38
rs778620013 Chr.4: 174262872-174262872 on Build GRCh38
rs749582981 Chr.4: 174262923-174262923 on Build GRCh38
rs756623069 Chr.4: 174262852-174262853 on Build GRCh38
rs753501050 Chr.4: 174262858-174262858 on Build GRCh38
rs375026512 Chr.4: 174262816-174262816 on Build GRCh38
rs138114332 Chr.4: 174262795-174262795 on Build GRCh38

View Full Product Details