Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7241266-7241507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABARAP
Entrez Gene ID
11337
Gene Name
GABA type A receptor-associated protein
Gene Aliases
ATG8A, GABARAP-a, MM46
UniGene
Hs.647421
Gene Chromosome Location
Chr.17: 7240419-7242434 on Build GRCh38
Transcripts
NM_007278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
707386 c.286C>A p.Q96K Chr.17: 7241344-7241344 on Build GRCh38
6242070 c.269C>A p.T90K Chr.17: 7241361-7241361 on Build GRCh38
1196094 c.238G>A p.V80I Chr.17: 7241392-7241392 on Build GRCh38
7145053 c.247G>A p.V83I Chr.17: 7241383-7241383 on Build GRCh38
1479945 c.186C>T p.F62F Chr.17: 7241444-7241444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367707073 Chr.17: 7241412-7241412 on Build GRCh38
rs765360232 Chr.17: 7241438-7241438 on Build GRCh38
rs372747001 Chr.17: 7241425-7241425 on Build GRCh38
rs773942051 Chr.17: 7241373-7241373 on Build GRCh38
rs201573431 Chr.17: 7241471-7241471 on Build GRCh38
rs376821360 Chr.17: 7241447-7241447 on Build GRCh38
rs755301333 Chr.17: 7241478-7241478 on Build GRCh38
rs1036071537 Chr.17: 7241336-7241336 on Build GRCh38
rs1048914801 Chr.17: 7241290-7241290 on Build GRCh38
rs762453489 Chr.17: 7241345-7241345 on Build GRCh38
rs887598092 Chr.17: 7241316-7241316 on Build GRCh38
rs747257323 Chr.17: 7241475-7241475 on Build GRCh38
rs139783498 Chr.17: 7241332-7241332 on Build GRCh38
rs762344521 Chr.17: 7241329-7241329 on Build GRCh38
rs774012076 Chr.17: 7241323-7241323 on Build GRCh38
rs371919454 Chr.17: 7241480-7241480 on Build GRCh38
rs756154103 Chr.17: 7241435-7241435 on Build GRCh38
rs747408184 Chr.17: 7241307-7241307 on Build GRCh38
rs758486874 Chr.17: 7241459-7241459 on Build GRCh38
rs766058157 Chr.17: 7241359-7241359 on Build GRCh38
rs112547582 Chr.17: 7241472-7241472 on Build GRCh38
rs111823949 Chr.17: 7241299-7241299 on Build GRCh38
rs760110442 Chr.17: 7241328-7241328 on Build GRCh38
rs759362721 Chr.17: 7241381-7241381 on Build GRCh38
rs948751665 Chr.17: 7241315-7241315 on Build GRCh38
rs772798713 Chr.17: 7241338-7241338 on Build GRCh38
rs777096591 Chr.17: 7241325-7241325 on Build GRCh38
rs1802217 Chr.17: 7241431-7241431 on Build GRCh38

View Full Product Details