Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 223002733-223002970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DISP1
Entrez Gene ID
84976
Gene Name
dispatched RND transporter family member 1
Gene Aliases
DISPA
UniGene
Hs.528817
Gene Chromosome Location
Chr.1: 222814514-223005995 on Build GRCh38
Transcripts
NM_032890.3 XM_005273335.2 XM_006711592.2 XM_006711594.3 XM_011510072.2 XM_011510073.2 XM_011510074.2 XM_011510075.2 XM_011510077.2 XM_017002611.1 XM_017002612.1 XM_017002613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4526247 c.1383G>A p.G461G Chr.1: 223002780-223002780 on Build GRCh38
1250127 c.1456G>A p.G486R Chr.1: 223002853-223002853 on Build GRCh38
5705232 c.1481_1484delGTTT p.L495fs*6 Chr.1: 223002878-223002881 on Build GRCh38
6644412 c.1441G>A p.V481M Chr.1: 223002838-223002838 on Build GRCh38
3484028 c.1430C>T p.S477F Chr.1: 223002827-223002827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762778909 Chr.1: 223002862-223002862 on Build GRCh38
rs112402042 Chr.1: 223002793-223002793 on Build GRCh38
rs779526244 Chr.1: 223002919-223002919 on Build GRCh38
rs1004724515 Chr.1: 223002930-223002930 on Build GRCh38
rs775124500 Chr.1: 223002853-223002853 on Build GRCh38
rs763697337 Chr.1: 223002867-223002867 on Build GRCh38
rs778079837 Chr.1: 223002931-223002931 on Build GRCh38
rs759386089 Chr.1: 223002851-223002851 on Build GRCh38
rs749772573 Chr.1: 223002771-223002771 on Build GRCh38
rs750019844 Chr.1: 223002914-223002914 on Build GRCh38
rs143019748 Chr.1: 223002756-223002756 on Build GRCh38
rs374203038 Chr.1: 223002837-223002837 on Build GRCh38
rs561363825 Chr.1: 223002917-223002917 on Build GRCh38
rs368376610 Chr.1: 223002939-223002939 on Build GRCh38
rs761256454 Chr.1: 223002907-223002907 on Build GRCh38
rs779142191 Chr.1: 223002827-223002827 on Build GRCh38
rs752961647 Chr.1: 223002922-223002922 on Build GRCh38
rs995199909 Chr.1: 223002841-223002841 on Build GRCh38
rs769827053 Chr.1: 223002852-223002852 on Build GRCh38
rs560069156 Chr.1: 223002834-223002834 on Build GRCh38
rs138673928 Chr.1: 223002830-223002830 on Build GRCh38
rs758853373 Chr.1: 223002928-223002928 on Build GRCh38
rs61738819 Chr.1: 223002933-223002933 on Build GRCh38
rs755303223 Chr.1: 223002797-223002797 on Build GRCh38
rs751236323 Chr.1: 223002885-223002885 on Build GRCh38
rs200592414 Chr.1: 223002803-223002803 on Build GRCh38
rs977342304 Chr.1: 223002836-223002836 on Build GRCh38
rs746007047 Chr.1: 223002946-223002946 on Build GRCh38
rs766527402 Chr.1: 223002798-223002798 on Build GRCh38
rs771148074 Chr.1: 223002844-223002844 on Build GRCh38
rs539816731 Chr.1: 223002913-223002913 on Build GRCh38
rs776561441 Chr.1: 223002847-223002847 on Build GRCh38
rs371983177 Chr.1: 223002838-223002838 on Build GRCh38
rs747235155 Chr.1: 223002932-223002932 on Build GRCh38

View Full Product Details