Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99710675-99710948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A7
Entrez Gene ID
1551
Gene Name
cytochrome P450 family 3 subfamily A member 7
Gene Aliases
CP37, CYPIIIA7, P-450(HFL33), P-450111A7, P450-HFLA
UniGene
Hs.111944
Gene Chromosome Location
Chr.7: 99705037-99735200 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Gene Symbol
CYP3A7-CYP3A51P
Entrez Gene ID
100861540
Gene Name
CYP3A7-CYP3A51P readthrough
Gene Aliases
CYP3A7-3AP1, CYP3A7-CYP3AP1, CYP3A7.1L
UniGene
Gene Chromosome Location
Chr.7: 99684679-99735200 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Gene Symbol
ZSCAN25
Entrez Gene ID
221785
Gene Name
zinc finger and SCAN domain containing 25
Gene Aliases
ZNF498
UniGene
Hs.446297
Gene Chromosome Location
Chr.7: 99616600-99738689 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232123 c.958G>A p.E320K Chr.7: 99710800-99710800 on Build GRCh38
3833630 c.971A>C p.H324P Chr.7: 99710787-99710787 on Build GRCh38
1453306 c.994C>T p.Q332* Chr.7: 99710764-99710764 on Build GRCh38
3642928 c.922G>A p.E308K Chr.7: 99710836-99710836 on Build GRCh38
6484467 c.886A>T p.M296L Chr.7: 99710872-99710872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768135942 Chr.7: 99710736-99710736 on Build GRCh38
rs772436106 Chr.7: 99710841-99710841 on Build GRCh38
rs1046639579 Chr.7: 99710785-99710785 on Build GRCh38
rs900348837 Chr.7: 99710904-99710904 on Build GRCh38
rs752990102 Chr.7: 99710740-99710740 on Build GRCh38
rs949527504 Chr.7: 99710783-99710783 on Build GRCh38
rs777212283 Chr.7: 99710767-99710767 on Build GRCh38
rs780092156 Chr.7: 99710794-99710794 on Build GRCh38
rs61737520 Chr.7: 99710829-99710829 on Build GRCh38
rs749464476 Chr.7: 99710828-99710828 on Build GRCh38
rs977826557 Chr.7: 99710697-99710697 on Build GRCh38
rs751495747 Chr.7: 99710926-99710926 on Build GRCh38
rs774504773 Chr.7: 99710726-99710726 on Build GRCh38
rs759894645 Chr.7: 99710729-99710729 on Build GRCh38
rs770122125 Chr.7: 99710717-99710717 on Build GRCh38
rs781158313 Chr.7: 99710809-99710809 on Build GRCh38
rs1040166922 Chr.7: 99710890-99710890 on Build GRCh38
rs756498783 Chr.7: 99710896-99710896 on Build GRCh38
rs111710676 Chr.7: 99710709-99710711 on Build GRCh38
rs918800257 Chr.7: 99710814-99710814 on Build GRCh38
rs749343393 Chr.7: 99710697-99710697 on Build GRCh38
rs763703301 Chr.7: 99710748-99710748 on Build GRCh38
rs370580138 Chr.7: 99710910-99710910 on Build GRCh38
rs140404982 Chr.7: 99710907-99710907 on Build GRCh38
rs754782383 Chr.7: 99710804-99710804 on Build GRCh38
rs45536632 Chr.7: 99710721-99710721 on Build GRCh38
rs758259550 Chr.7: 99710774-99710774 on Build GRCh38
rs781002724 Chr.7: 99710810-99710810 on Build GRCh38
rs370772742 Chr.7: 99710701-99710701 on Build GRCh38
rs749883684 Chr.7: 99710889-99710889 on Build GRCh38
rs764390751 Chr.7: 99710853-99710853 on Build GRCh38
rs916774586 Chr.7: 99710782-99710782 on Build GRCh38
rs778224174 Chr.7: 99710754-99710754 on Build GRCh38
rs377206089 Chr.7: 99710835-99710835 on Build GRCh38
rs765003638 Chr.7: 99710885-99710885 on Build GRCh38
rs372734706 Chr.7: 99710746-99710746 on Build GRCh38
rs746884273 Chr.7: 99710715-99710715 on Build GRCh38
rs776103784 Chr.7: 99710848-99710848 on Build GRCh38
rs779644303 Chr.7: 99710698-99710698 on Build GRCh38
rs199557001 Chr.7: 99710699-99710699 on Build GRCh38
rs772071749 Chr.7: 99710708-99710710 on Build GRCh38
rs45592732 Chr.7: 99710722-99710722 on Build GRCh38
rs773275521 Chr.7: 99710747-99710748 on Build GRCh38
rs776990396 Chr.7: 99710875-99710875 on Build GRCh38
rs41279863 Chr.7: 99710851-99710851 on Build GRCh38
rs199558694 Chr.7: 99710868-99710868 on Build GRCh38
rs147031398 Chr.7: 99710758-99710758 on Build GRCh38
rs746773235 Chr.7: 99710795-99710795 on Build GRCh38
rs746277130 Chr.7: 99710876-99710876 on Build GRCh38
rs756922005 Chr.7: 99710749-99710749 on Build GRCh38

View Full Product Details