Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15833477-15833710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM188A
Entrez Gene ID
80013
Gene Name
family with sequence similarity 188 member A
Gene Aliases
C10orf97, CARP, DERP5, MST126, MSTP126, my042
UniGene
Hs.158870
Gene Chromosome Location
Chr.10: 15778169-15860533 on Build GRCh38
Transcripts
NM_001318330.1 NM_024948.3 XM_005252600.2 XM_005252602.2 XM_006717508.2 XM_011519690.2 XM_011519691.2 XM_011519693.1 XM_011519694.1 XM_017016671.1 XM_017016672.1 XM_017016673.1 XM_017016674.1 XM_017016675.1 XM_017016676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
465494 c.678G>A p.T226T Chr.10: 15833682-15833682 on Build GRCh38
5443573 c.680delG p.G227fs*17 Chr.10: 15833680-15833680 on Build GRCh38
3978319 c.671T>A p.L224Q Chr.10: 15833689-15833689 on Build GRCh38
4426733 c.240A>G p.G80G Chr.10: 15833679-15833679 on Build GRCh38
1297141 c.728T>C p.M243T Chr.10: 15833632-15833632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775319044 Chr.10: 15833625-15833625 on Build GRCh38
rs761405837 Chr.10: 15833666-15833666 on Build GRCh38
rs950147903 Chr.10: 15833573-15833573 on Build GRCh38
rs893065212 Chr.10: 15833653-15833653 on Build GRCh38
rs777971741 Chr.10: 15833583-15833583 on Build GRCh38
rs745698436 Chr.10: 15833547-15833547 on Build GRCh38
rs912465541 Chr.10: 15833544-15833544 on Build GRCh38
rs760037047 Chr.10: 15833680-15833680 on Build GRCh38
rs369452204 Chr.10: 15833631-15833631 on Build GRCh38
rs1045753737 Chr.10: 15833592-15833592 on Build GRCh38
rs550826094 Chr.10: 15833603-15833603 on Build GRCh38
rs745870397 Chr.10: 15833611-15833611 on Build GRCh38
rs150118445 Chr.10: 15833661-15833661 on Build GRCh38
rs564068812 Chr.10: 15833506-15833506 on Build GRCh38
rs767177975 Chr.10: 15833677-15833677 on Build GRCh38
rs749102722 Chr.10: 15833657-15833657 on Build GRCh38
rs866547768 Chr.10: 15833641-15833641 on Build GRCh38
rs774204974 Chr.10: 15833662-15833662 on Build GRCh38
rs910158201 Chr.10: 15833529-15833529 on Build GRCh38
rs941604468 Chr.10: 15833543-15833543 on Build GRCh38
rs565190843 Chr.10: 15833553-15833553 on Build GRCh38
rs769643453 Chr.10: 15833621-15833621 on Build GRCh38
rs776980502 Chr.10: 15833679-15833679 on Build GRCh38
rs747029395 Chr.10: 15833598-15833598 on Build GRCh38
rs201352822 Chr.10: 15833683-15833683 on Build GRCh38
rs185974010 Chr.10: 15833601-15833601 on Build GRCh38
rs903657477 Chr.10: 15833660-15833660 on Build GRCh38
rs753025159 Chr.10: 15833581-15833581 on Build GRCh38
rs1042334762 Chr.10: 15833617-15833617 on Build GRCh38
rs563431292 Chr.10: 15833682-15833682 on Build GRCh38

View Full Product Details