Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 71516045-71516308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYSF
Entrez Gene ID
8291
Gene Name
dysferlin
Gene Aliases
FER1L1, LGMD2B, MMD1
UniGene
Hs.252180
Gene Chromosome Location
Chr.2: 71453155-71686763 on Build GRCh38
Transcripts
NM_001130455.1 NM_001130976.1 NM_001130977.1 NM_001130978.1 NM_001130979.1 NM_001130980.1 NM_001130981.1 NM_001130982.1 NM_001130983.1 NM_001130984.1 NM_001130985.1 NM_001130986.1 NM_001130987.1 NM_003494.3 XM_005264584.4 XM_005264585.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
721916 c.823G>T p.G275W Chr.2: 71516210-71516210 on Build GRCh38
1483236 c.926T>G p.L309R Chr.2: 71516217-71516217 on Build GRCh38
4095472 c.847T>A p.F283I Chr.2: 71516234-71516234 on Build GRCh38
1022661 c.846C>G p.I282M Chr.2: 71516233-71516233 on Build GRCh38
4095468 c.793A>G p.T265A Chr.2: 71516180-71516180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551384644 Chr.2: 71516233-71516233 on Build GRCh38
rs370936168 Chr.2: 71516262-71516262 on Build GRCh38
rs774059957 Chr.2: 71516193-71516193 on Build GRCh38
rs147675576 Chr.2: 71516236-71516236 on Build GRCh38
rs115184725 Chr.2: 71516146-71516146 on Build GRCh38
rs111581279 Chr.2: 71516227-71516227 on Build GRCh38
rs529400653 Chr.2: 71516207-71516207 on Build GRCh38
rs750376064 Chr.2: 71516282-71516282 on Build GRCh38
rs201128463 Chr.2: 71516153-71516153 on Build GRCh38
rs11903960 Chr.2: 71516246-71516246 on Build GRCh38
rs988491095 Chr.2: 71516183-71516183 on Build GRCh38
rs533297432 Chr.2: 71516126-71516127 on Build GRCh38
rs139753518 Chr.2: 71516143-71516143 on Build GRCh38
rs1004869329 Chr.2: 71516128-71516128 on Build GRCh38
rs934428561 Chr.2: 71516178-71516178 on Build GRCh38
rs372952394 Chr.2: 71516241-71516241 on Build GRCh38
rs13417592 Chr.2: 71516076-71516076 on Build GRCh38
rs893232964 Chr.2: 71516104-71516104 on Build GRCh38
rs74384941 Chr.2: 71516281-71516281 on Build GRCh38
rs975366663 Chr.2: 71516151-71516151 on Build GRCh38
rs771253287 Chr.2: 71516242-71516242 on Build GRCh38
rs374114774 Chr.2: 71516287-71516287 on Build GRCh38
rs889590190 Chr.2: 71516127-71516127 on Build GRCh38
rs765338016 Chr.2: 71516134-71516134 on Build GRCh38
rs916319528 Chr.2: 71516197-71516197 on Build GRCh38
rs768207094 Chr.2: 71516187-71516187 on Build GRCh38
rs755059821 Chr.2: 71516131-71516131 on Build GRCh38
rs1002728050 Chr.2: 71516099-71516099 on Build GRCh38
rs954851578 Chr.2: 71516154-71516154 on Build GRCh38
rs865798392 Chr.2: 71516191-71516191 on Build GRCh38
rs934694477 Chr.2: 71516069-71516069 on Build GRCh38
rs377156563 Chr.2: 71516155-71516155 on Build GRCh38
rs786200898 Chr.2: 71516243-71516243 on Build GRCh38
rs369502694 Chr.2: 71516149-71516149 on Build GRCh38
rs114384446 Chr.2: 71516271-71516271 on Build GRCh38
rs886043967 Chr.2: 71516229-71516229 on Build GRCh38
rs886554317 Chr.2: 71516123-71516123 on Build GRCh38
rs752788962 Chr.2: 71516270-71516270 on Build GRCh38
rs886042590 Chr.2: 71516244-71516244 on Build GRCh38
rs759746992 Chr.2: 71516247-71516247 on Build GRCh38
rs751491934 Chr.2: 71516140-71516140 on Build GRCh38
rs912300172 Chr.2: 71516173-71516173 on Build GRCh38
rs187954641 Chr.2: 71516111-71516111 on Build GRCh38
rs753807885 Chr.2: 71516129-71516129 on Build GRCh38
rs200969236 Chr.2: 71516274-71516274 on Build GRCh38
rs772708040 Chr.2: 71516224-71516224 on Build GRCh38
rs532691220 Chr.2: 71516232-71516232 on Build GRCh38
rs370223980 Chr.2: 71516174-71516174 on Build GRCh38
rs748604967 Chr.2: 71516203-71516203 on Build GRCh38
rs905658646 Chr.2: 71516092-71516092 on Build GRCh38
rs398123806 Chr.2: 71516184-71516184 on Build GRCh38
rs28500662 Chr.2: 71516112-71516112 on Build GRCh38

View Full Product Details