Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16045485-16045746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCNKB
Entrez Gene ID
1188
Gene Name
chloride voltage-gated channel Kb
Gene Aliases
CLCKB, ClC-K2, ClC-Kb
UniGene
Hs.352243
Gene Chromosome Location
Chr.1: 16043736-16057326 on Build GRCh38
Transcripts
NM_000085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361441 c.134G>T p.R45L Chr.1: 16045591-16045591 on Build GRCh38
5844337 c.166C>A p.L56I Chr.1: 16045623-16045623 on Build GRCh38
4473434 c.184C>T p.L62L Chr.1: 16045641-16045641 on Build GRCh38
5140814 c.147C>G p.D49E Chr.1: 16045604-16045604 on Build GRCh38
1248385 c.159G>A p.L53L Chr.1: 16045616-16045616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571646834 Chr.1: 16045622-16045622 on Build GRCh38
rs751569887 Chr.1: 16045656-16045656 on Build GRCh38
rs201954126 Chr.1: 16045698-16045698 on Build GRCh38
rs139676842 Chr.1: 16045684-16045684 on Build GRCh38
rs757241954 Chr.1: 16045666-16045666 on Build GRCh38
rs769604665 Chr.1: 16045599-16045599 on Build GRCh38
rs148698605 Chr.1: 16045584-16045584 on Build GRCh38
rs771497331 Chr.1: 16045699-16045699 on Build GRCh38
rs538083063 Chr.1: 16045544-16045544 on Build GRCh38
rs138768237 Chr.1: 16045562-16045562 on Build GRCh38
rs147391626 Chr.1: 16045568-16045568 on Build GRCh38
rs777646118 Chr.1: 16045520-16045520 on Build GRCh38
rs778531035 Chr.1: 16045693-16045693 on Build GRCh38
rs777700978 Chr.1: 16045588-16045588 on Build GRCh38
rs746245249 Chr.1: 16045722-16045722 on Build GRCh38
rs144043939 Chr.1: 16045629-16045629 on Build GRCh38
rs761281486 Chr.1: 16045633-16045633 on Build GRCh38
rs551946850 Chr.1: 16045591-16045591 on Build GRCh38
rs553995820 Chr.1: 16045653-16045653 on Build GRCh38
rs764275060 Chr.1: 16045655-16045655 on Build GRCh38
rs767320052 Chr.1: 16045667-16045667 on Build GRCh38
rs750240299 Chr.1: 16045677-16045677 on Build GRCh38
rs979233227 Chr.1: 16045623-16045623 on Build GRCh38
rs759852388 Chr.1: 16045640-16045640 on Build GRCh38
rs143673726 Chr.1: 16045574-16045574 on Build GRCh38
rs745575993 Chr.1: 16045532-16045532 on Build GRCh38
rs777073739 Chr.1: 16045639-16045639 on Build GRCh38
rs6604910 Chr.1: 16045508-16045508 on Build GRCh38
rs528798067 Chr.1: 16045626-16045626 on Build GRCh38
rs755918269 Chr.1: 16045679-16045679 on Build GRCh38
rs763148018 Chr.1: 16045654-16045654 on Build GRCh38
rs759799517 Chr.1: 16045579-16045579 on Build GRCh38
rs771316846 Chr.1: 16045636-16045636 on Build GRCh38
rs150723240 Chr.1: 16045628-16045628 on Build GRCh38
rs776933851 Chr.1: 16045713-16045713 on Build GRCh38
rs758352654 Chr.1: 16045518-16045518 on Build GRCh38
rs375301415 Chr.1: 16045569-16045569 on Build GRCh38
rs569318749 Chr.1: 16045535-16045535 on Build GRCh38
rs762515275 Chr.1: 16045558-16045558 on Build GRCh38
rs367552783 Chr.1: 16045590-16045590 on Build GRCh38
rs370273137 Chr.1: 16045598-16045598 on Build GRCh38
rs370985865 Chr.1: 16045683-16045683 on Build GRCh38
rs770701845 Chr.1: 16045521-16045521 on Build GRCh38
rs780845638 Chr.1: 16045529-16045529 on Build GRCh38
rs754143576 Chr.1: 16045507-16045507 on Build GRCh38
rs781091568 Chr.1: 16045597-16045597 on Build GRCh38
rs778941895 Chr.1: 16045510-16045510 on Build GRCh38
rs765392974 Chr.1: 16045583-16045583 on Build GRCh38

View Full Product Details