Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123768508-123768779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM91A1
Entrez Gene ID
157769
Gene Name
family with sequence similarity 91 member A1
Gene Aliases
-
UniGene
Hs.459174
Gene Chromosome Location
Chr.8: 123768439-123815452 on Build GRCh38
Transcripts
NM_001317917.1 NM_001317918.1 NM_144963.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1755555 c.8T>G p.I3R Chr.8: 123768710-123768710 on Build GRCh38
6289677 c.1A>G p.M1V Chr.8: 123768703-123768703 on Build GRCh38
1095810 c.59C>T p.A20V Chr.8: 123768761-123768761 on Build GRCh38
5647477 c.66G>C p.V22V Chr.8: 123768768-123768768 on Build GRCh38
1095809 c.17A>T p.E6V Chr.8: 123768719-123768719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966707133 Chr.8: 123768658-123768659 on Build GRCh38
rs1054591760 Chr.8: 123768533-123768533 on Build GRCh38
rs181046952 Chr.8: 123768618-123768618 on Build GRCh38
rs972192090 Chr.8: 123768645-123768645 on Build GRCh38
rs755290849 Chr.8: 123768759-123768759 on Build GRCh38
rs765116609 Chr.8: 123768715-123768717 on Build GRCh38
rs12334564 Chr.8: 123768584-123768584 on Build GRCh38
rs998629667 Chr.8: 123768532-123768532 on Build GRCh38
rs775303795 Chr.8: 123768660-123768660 on Build GRCh38
rs749018169 Chr.8: 123768676-123768676 on Build GRCh38
rs996880599 Chr.8: 123768569-123768569 on Build GRCh38
rs1017844510 Chr.8: 123768644-123768644 on Build GRCh38
rs576218080 Chr.8: 123768550-123768550 on Build GRCh38
rs761599272 Chr.8: 123768731-123768731 on Build GRCh38
rs546136372 Chr.8: 123768633-123768633 on Build GRCh38
rs767449243 Chr.8: 123768732-123768732 on Build GRCh38
rs142309473 Chr.8: 123768581-123768581 on Build GRCh38
rs747380687 Chr.8: 123768652-123768652 on Build GRCh38
rs574564231 Chr.8: 123768575-123768575 on Build GRCh38
rs890622782 Chr.8: 123768605-123768605 on Build GRCh38
rs982184377 Chr.8: 123768681-123768681 on Build GRCh38
rs773969528 Chr.8: 123768709-123768709 on Build GRCh38
rs560007370 Chr.8: 123768658-123768658 on Build GRCh38
rs760488117 Chr.8: 123768743-123768743 on Build GRCh38
rs368997819 Chr.8: 123768695-123768695 on Build GRCh38
rs961164557 Chr.8: 123768626-123768626 on Build GRCh38
rs1037081373 Chr.8: 123768560-123768560 on Build GRCh38
rs1029403761 Chr.8: 123768599-123768599 on Build GRCh38
rs772955315 Chr.8: 123768739-123768739 on Build GRCh38
rs892804157 Chr.8: 123768568-123768568 on Build GRCh38
rs937841472 Chr.8: 123768710-123768710 on Build GRCh38
rs766214631 Chr.8: 123768752-123768752 on Build GRCh38
rs758956866 Chr.8: 123768573-123768573 on Build GRCh38
rs753980063 Chr.8: 123768758-123768758 on Build GRCh38

View Full Product Details