Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124756140-124756401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESAM
Entrez Gene ID
90952
Gene Name
endothelial cell adhesion molecule
Gene Aliases
W117m
UniGene
Hs.173840
Gene Chromosome Location
Chr.11: 124753123-124762327 on Build GRCh38
Transcripts
NM_138961.2 XM_005271720.4 XM_011543060.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6296721 c.607+10C>A p.? Chr.11: 124756197-124756197 on Build GRCh38
686575 c.562G>T p.D188Y Chr.11: 124756252-124756252 on Build GRCh38
1352298 c.514C>A p.L172M Chr.11: 124756300-124756300 on Build GRCh38
3868728 c.455C>T p.P152L Chr.11: 124756359-124756359 on Build GRCh38
4018961 c.500G>A p.G167E Chr.11: 124756314-124756314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767004846 Chr.11: 124756183-124756183 on Build GRCh38
rs368969134 Chr.11: 124756200-124756200 on Build GRCh38
rs951616294 Chr.11: 124756373-124756373 on Build GRCh38
rs754064863 Chr.11: 124756323-124756323 on Build GRCh38
rs61020077 Chr.11: 124756194-124756194 on Build GRCh38
rs764969890 Chr.11: 124756226-124756226 on Build GRCh38
rs748308260 Chr.11: 124756243-124756243 on Build GRCh38
rs773710813 Chr.11: 124756163-124756163 on Build GRCh38
rs377500728 Chr.11: 124756339-124756339 on Build GRCh38
rs201583234 Chr.11: 124756237-124756237 on Build GRCh38
rs750322401 Chr.11: 124756228-124756228 on Build GRCh38
rs749731089 Chr.11: 124756166-124756166 on Build GRCh38
rs747543095 Chr.11: 124756374-124756374 on Build GRCh38
rs569407653 Chr.11: 124756306-124756306 on Build GRCh38
rs765020444 Chr.11: 124756300-124756300 on Build GRCh38
rs763791454 Chr.11: 124756197-124756197 on Build GRCh38
rs757435206 Chr.11: 124756338-124756338 on Build GRCh38
rs759993093 Chr.11: 124756175-124756175 on Build GRCh38
rs752805703 Chr.11: 124756311-124756311 on Build GRCh38
rs200015552 Chr.11: 124756248-124756248 on Build GRCh38
rs372960582 Chr.11: 124756220-124756220 on Build GRCh38
rs375327056 Chr.11: 124756249-124756249 on Build GRCh38
rs781270174 Chr.11: 124756238-124756238 on Build GRCh38
rs771286783 Chr.11: 124756252-124756252 on Build GRCh38
rs756303624 Chr.11: 124756313-124756313 on Build GRCh38
rs746274223 Chr.11: 124756351-124756351 on Build GRCh38
rs202017012 Chr.11: 124756195-124756195 on Build GRCh38
rs779588823 Chr.11: 124756266-124756266 on Build GRCh38
rs1040464556 Chr.11: 124756240-124756240 on Build GRCh38
rs776223057 Chr.11: 124756293-124756293 on Build GRCh38
rs778032762 Chr.11: 124756319-124756319 on Build GRCh38
rs768367886 Chr.11: 124756292-124756292 on Build GRCh38
rs774984719 Chr.11: 124756191-124756191 on Build GRCh38
rs761390500 Chr.11: 124756299-124756299 on Build GRCh38
rs771322750 Chr.11: 124756169-124756169 on Build GRCh38
rs779168741 Chr.11: 124756261-124756261 on Build GRCh38
rs149527591 Chr.11: 124756307-124756307 on Build GRCh38
rs199848606 Chr.11: 124756304-124756304 on Build GRCh38
rs985056436 Chr.11: 124756192-124756192 on Build GRCh38
rs780575270 Chr.11: 124756353-124756353 on Build GRCh38
rs772521572 Chr.11: 124756270-124756270 on Build GRCh38
rs774964649 Chr.11: 124756172-124756172 on Build GRCh38
rs768101946 Chr.11: 124756376-124756376 on Build GRCh38
rs139265422 Chr.11: 124756271-124756271 on Build GRCh38
rs757081410 Chr.11: 124756202-124756202 on Build GRCh38
rs768263465 Chr.11: 124756275-124756275 on Build GRCh38
rs903686914 Chr.11: 124756264-124756264 on Build GRCh38
rs772608506 Chr.11: 124756352-124756352 on Build GRCh38

View Full Product Details