Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32488191-32488442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5R1
Entrez Gene ID
8851
Gene Name
cyclin dependent kinase 5 regulatory subunit 1
Gene Aliases
CDK5P35, CDK5R, NCK5A, p23, p25, p35, p35nck5a
UniGene
Hs.500015
Gene Chromosome Location
Chr.17: 32485281-32491256 on Build GRCh38
Transcripts
NM_003885.2 XM_011525438.2 XM_017025281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3983368 c.800T>G p.L267R Chr.17: 32488420-32488420 on Build GRCh38
6456038 c.586A>G p.I196V Chr.17: 32488206-32488206 on Build GRCh38
6331294 c.773G>T p.W258L Chr.17: 32488393-32488393 on Build GRCh38
977630 c.795C>T p.I265I Chr.17: 32488415-32488415 on Build GRCh38
4065416 c.590C>T p.T197M Chr.17: 32488210-32488210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753521866 Chr.17: 32488229-32488229 on Build GRCh38
rs764730659 Chr.17: 32488286-32488286 on Build GRCh38
rs765126369 Chr.17: 32488316-32488316 on Build GRCh38
rs764319776 Chr.17: 32488211-32488211 on Build GRCh38
rs775846634 Chr.17: 32488278-32488278 on Build GRCh38
rs780702599 Chr.17: 32488349-32488349 on Build GRCh38
rs919470726 Chr.17: 32488376-32488376 on Build GRCh38
rs776358749 Chr.17: 32488265-32488265 on Build GRCh38
rs745590491 Chr.17: 32488352-32488352 on Build GRCh38
rs754718606 Chr.17: 32488312-32488312 on Build GRCh38
rs949720216 Chr.17: 32488343-32488343 on Build GRCh38
rs34511797 Chr.17: 32488418-32488418 on Build GRCh38
rs761983951 Chr.17: 32488408-32488408 on Build GRCh38
rs777302939 Chr.17: 32488256-32488256 on Build GRCh38
rs777707085 Chr.17: 32488345-32488345 on Build GRCh38
rs758287417 Chr.17: 32488337-32488337 on Build GRCh38
rs1039134075 Chr.17: 32488391-32488391 on Build GRCh38
rs574969901 Chr.17: 32488247-32488247 on Build GRCh38
rs763595725 Chr.17: 32488280-32488280 on Build GRCh38
rs781499965 Chr.17: 32488223-32488223 on Build GRCh38
rs778714379 Chr.17: 32488244-32488244 on Build GRCh38
rs867757984 Chr.17: 32488215-32488215 on Build GRCh38
rs17852413 Chr.17: 32488263-32488263 on Build GRCh38
rs757878520 Chr.17: 32488252-32488252 on Build GRCh38
rs1032779285 Chr.17: 32488210-32488210 on Build GRCh38
rs532658080 Chr.17: 32488311-32488311 on Build GRCh38
rs770024512 Chr.17: 32488369-32488369 on Build GRCh38
rs770518849 Chr.17: 32488262-32488262 on Build GRCh38
rs767696040 Chr.17: 32488410-32488410 on Build GRCh38
rs775959493 Chr.17: 32488385-32488385 on Build GRCh38
rs754811246 Chr.17: 32488232-32488232 on Build GRCh38
rs929494932 Chr.17: 32488377-32488377 on Build GRCh38
rs757514550 Chr.17: 32488214-32488214 on Build GRCh38
rs757908913 Chr.17: 32488355-32488355 on Build GRCh38
rs752596930 Chr.17: 32488317-32488317 on Build GRCh38
rs767573470 Chr.17: 32488308-32488308 on Build GRCh38
rs749861167 Chr.17: 32488239-32488239 on Build GRCh38
rs746549493 Chr.17: 32488259-32488259 on Build GRCh38
rs560668827 Chr.17: 32488292-32488292 on Build GRCh38
rs943457059 Chr.17: 32488388-32488388 on Build GRCh38
rs762002030 Chr.17: 32488296-32488296 on Build GRCh38
rs775006795 Chr.17: 32488398-32488398 on Build GRCh38
rs143370438 Chr.17: 32488274-32488274 on Build GRCh38
rs201269434 Chr.17: 32488381-32488381 on Build GRCh38
rs746463658 Chr.17: 32488346-32488346 on Build GRCh38
rs199565053 Chr.17: 32488387-32488387 on Build GRCh38

View Full Product Details