Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158170090-158170363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM198B
Entrez Gene ID
51313
Gene Name
family with sequence similarity 198 member B
Gene Aliases
AD021, AD036, C4orf18
UniGene
Hs.567498
Gene Chromosome Location
Chr.4: 158124580-158173050 on Build GRCh38
Transcripts
NM_001031700.2 NM_001128424.1 NM_016613.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1684011 c.911-3_911-1delCAG p.? Chr.4: 158170348-158170350 on Build GRCh38
6047307 c.998T>G p.I333S Chr.4: 158170260-158170260 on Build GRCh38
6441969 c.982_983CC>TT p.P328L Chr.4: 158170275-158170276 on Build GRCh38
5699892 c.970G>C p.D324H Chr.4: 158170288-158170288 on Build GRCh38
4584878 c.968A>G p.E323G Chr.4: 158170290-158170290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755256963 Chr.4: 158170310-158170310 on Build GRCh38
rs758505182 Chr.4: 158170164-158170164 on Build GRCh38
rs749301292 Chr.4: 158170335-158170335 on Build GRCh38
rs748759943 Chr.4: 158170335-158170335 on Build GRCh38
rs148372081 Chr.4: 158170266-158170266 on Build GRCh38
rs780702139 Chr.4: 158170337-158170337 on Build GRCh38
rs780402444 Chr.4: 158170297-158170297 on Build GRCh38
rs576932247 Chr.4: 158170217-158170217 on Build GRCh38
rs754864207 Chr.4: 158170334-158170335 on Build GRCh38
rs376987062 Chr.4: 158170219-158170219 on Build GRCh38
rs780457171 Chr.4: 158170169-158170169 on Build GRCh38
rs548251612 Chr.4: 158170129-158170129 on Build GRCh38
rs146894527 Chr.4: 158170254-158170254 on Build GRCh38
rs764783404 Chr.4: 158170285-158170285 on Build GRCh38
rs369885932 Chr.4: 158170206-158170206 on Build GRCh38
rs201127725 Chr.4: 158170258-158170258 on Build GRCh38
rs758703457 Chr.4: 158170197-158170197 on Build GRCh38
rs755764059 Chr.4: 158170313-158170313 on Build GRCh38
rs539644643 Chr.4: 158170231-158170231 on Build GRCh38
rs758504988 Chr.4: 158170156-158170157 on Build GRCh38
rs1049223283 Chr.4: 158170153-158170153 on Build GRCh38
rs758984039 Chr.4: 158170294-158170294 on Build GRCh38
rs748307483 Chr.4: 158170321-158170321 on Build GRCh38
rs769795651 Chr.4: 158170213-158170213 on Build GRCh38
rs749941386 Chr.4: 158170289-158170289 on Build GRCh38
rs373324742 Chr.4: 158170238-158170238 on Build GRCh38
rs779597366 Chr.4: 158170196-158170196 on Build GRCh38
rs150507892 Chr.4: 158170272-158170272 on Build GRCh38
rs762316396 Chr.4: 158170235-158170235 on Build GRCh38
rs922612444 Chr.4: 158170135-158170135 on Build GRCh38
rs145106513 Chr.4: 158170270-158170270 on Build GRCh38
rs555377085 Chr.4: 158170306-158170306 on Build GRCh38
rs368072438 Chr.4: 158170202-158170202 on Build GRCh38
rs976646460 Chr.4: 158170120-158170120 on Build GRCh38
rs930779065 Chr.4: 158170132-158170132 on Build GRCh38
rs781394185 Chr.4: 158170320-158170320 on Build GRCh38
rs372519767 Chr.4: 158170207-158170207 on Build GRCh38
rs370382961 Chr.4: 158170225-158170225 on Build GRCh38
rs750509992 Chr.4: 158170147-158170147 on Build GRCh38
rs868577681 Chr.4: 158170179-158170179 on Build GRCh38
rs759571582 Chr.4: 158170222-158170222 on Build GRCh38
rs376585720 Chr.4: 158170327-158170327 on Build GRCh38
rs777512274 Chr.4: 158170335-158170337 on Build GRCh38
rs760367173 Chr.4: 158170252-158170252 on Build GRCh38
rs769987028 Chr.4: 158170324-158170324 on Build GRCh38
rs568159974 Chr.4: 158170154-158170154 on Build GRCh38
rs772098128 Chr.4: 158170338-158170338 on Build GRCh38

View Full Product Details