Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 411612-411859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC77
Entrez Gene ID
84318
Gene Name
coiled-coil domain containing 77
Gene Aliases
-
UniGene
Hs.631656
Gene Chromosome Location
Chr.12: 389350-442642 on Build GRCh38
Transcripts
NM_001130146.1 NM_001130147.1 NM_001130148.1 NM_032358.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
309923 c.70G>A p.V24I Chr.12: 411778-411778 on Build GRCh38
4629963 c.59G>A p.R20H Chr.12: 411767-411767 on Build GRCh38
3998831 c.73A>C p.S25R Chr.12: 411781-411781 on Build GRCh38
6866528 c.92G>T p.R31M Chr.12: 411800-411800 on Build GRCh38
1361479 c.143G>A p.R48H Chr.12: 411851-411851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774073292 Chr.12: 411777-411777 on Build GRCh38
rs191611852 Chr.12: 411706-411706 on Build GRCh38
rs984790017 Chr.12: 411711-411711 on Build GRCh38
rs762989082 Chr.12: 411770-411770 on Build GRCh38
rs144639108 Chr.12: 411831-411831 on Build GRCh38
rs754125267 Chr.12: 411782-411782 on Build GRCh38
rs929495998 Chr.12: 411763-411763 on Build GRCh38
rs770027750 Chr.12: 411698-411699 on Build GRCh38
rs781370354 Chr.12: 411815-411815 on Build GRCh38
rs201758521 Chr.12: 411748-411748 on Build GRCh38
rs778928477 Chr.12: 411715-411715 on Build GRCh38
rs750431707 Chr.12: 411792-411792 on Build GRCh38
rs772923533 Chr.12: 411823-411823 on Build GRCh38
rs4980895 Chr.12: 411781-411781 on Build GRCh38
rs771384665 Chr.12: 411767-411769 on Build GRCh38
rs765311307 Chr.12: 411791-411791 on Build GRCh38
rs753916412 Chr.12: 411708-411708 on Build GRCh38
rs140519509 Chr.12: 411707-411707 on Build GRCh38
rs1018379138 Chr.12: 411680-411680 on Build GRCh38
rs776322698 Chr.12: 411749-411749 on Build GRCh38
rs1034770408 Chr.12: 411836-411836 on Build GRCh38
rs371407974 Chr.12: 411743-411743 on Build GRCh38
rs772506727 Chr.12: 411742-411742 on Build GRCh38
rs949162193 Chr.12: 411761-411761 on Build GRCh38
rs747393530 Chr.12: 411747-411747 on Build GRCh38
rs759355080 Chr.12: 411839-411839 on Build GRCh38
rs979843928 Chr.12: 411691-411691 on Build GRCh38
rs117439611 Chr.12: 411778-411778 on Build GRCh38
rs562220233 Chr.12: 411719-411719 on Build GRCh38
rs769459378 Chr.12: 411762-411762 on Build GRCh38
rs998702563 Chr.12: 411824-411824 on Build GRCh38
rs747380154 Chr.12: 411807-411807 on Build GRCh38
rs761267852 Chr.12: 411753-411753 on Build GRCh38
rs1046135737 Chr.12: 411766-411766 on Build GRCh38
rs773461093 Chr.12: 411710-411711 on Build GRCh38
rs550926839 Chr.12: 411787-411787 on Build GRCh38
rs763155909 Chr.12: 411740-411740 on Build GRCh38
rs141015442 Chr.12: 411826-411826 on Build GRCh38
rs370768978 Chr.12: 411767-411767 on Build GRCh38
rs767508797 Chr.12: 411776-411776 on Build GRCh38
rs35234577 Chr.12: 411810-411810 on Build GRCh38
rs769504422 Chr.12: 411816-411816 on Build GRCh38
rs753250167 Chr.12: 411672-411672 on Build GRCh38
rs528763460 Chr.12: 411670-411670 on Build GRCh38
rs550645891 Chr.12: 411775-411775 on Build GRCh38
rs564756598 Chr.12: 411657-411657 on Build GRCh38
rs370744667 Chr.12: 411799-411799 on Build GRCh38
rs757298154 Chr.12: 411712-411712 on Build GRCh38

View Full Product Details