Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140672779-140673004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DND1
Entrez Gene ID
373863
Gene Name
DND microRNA-mediated repression inhibitor 1
Gene Aliases
RBMS4
UniGene
Hs.591262
Gene Chromosome Location
Chr.5: 140670796-140673586 on Build GRCh38
Transcripts
NM_194249.2 XM_005268469.2 XM_017009600.1

Gene Symbol
WDR55
Entrez Gene ID
54853
Gene Name
WD repeat domain 55
Gene Aliases
-
UniGene
Hs.286261
Gene Chromosome Location
Chr.5: 140664799-140674119 on Build GRCh38
Transcripts
NM_194249.2 XM_005268469.2 XM_017009600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416506 c.248G>T p.R83L Chr.5: 140672801-140672801 on Build GRCh38
6430586 c.236A>G p.Q79R Chr.5: 140672813-140672813 on Build GRCh38
5763309 c.218A>G p.Q73R Chr.5: 140672831-140672831 on Build GRCh38
5222898 c.165A>G p.P55P Chr.5: 140672884-140672884 on Build GRCh38
1130919 c.227C>T p.P76L Chr.5: 140672822-140672822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761795938 Chr.5: 140672857-140672857 on Build GRCh38
rs72800920 Chr.5: 140672822-140672822 on Build GRCh38
rs762997439 Chr.5: 140672884-140672884 on Build GRCh38
rs75826220 Chr.5: 140672845-140672845 on Build GRCh38
rs773335514 Chr.5: 140672883-140672883 on Build GRCh38
rs35269884 Chr.5: 140672803-140672803 on Build GRCh38
rs571658944 Chr.5: 140672925-140672925 on Build GRCh38
rs72800921 Chr.5: 140672839-140672839 on Build GRCh38
rs769657267 Chr.5: 140672859-140672859 on Build GRCh38
rs199742545 Chr.5: 140672944-140672944 on Build GRCh38
rs776601209 Chr.5: 140672847-140672847 on Build GRCh38
rs556683206 Chr.5: 140672802-140672802 on Build GRCh38
rs1047645431 Chr.5: 140672849-140672849 on Build GRCh38
rs766489935 Chr.5: 140672893-140672893 on Build GRCh38
rs866222509 Chr.5: 140672919-140672919 on Build GRCh38
rs1008857828 Chr.5: 140672979-140672979 on Build GRCh38
rs1018655673 Chr.5: 140672983-140672983 on Build GRCh38
rs759784951 Chr.5: 140672934-140672934 on Build GRCh38
rs778463388 Chr.5: 140672815-140672815 on Build GRCh38

View Full Product Details