Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 57550623-57550894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FECH
Entrez Gene ID
2235
Gene Name
ferrochelatase
Gene Aliases
EPP, FCE
UniGene
Hs.365365
Gene Chromosome Location
Chr.18: 57544841-57586737 on Build GRCh38
Transcripts
NM_000140.3 NM_001012515.2 XM_011525881.1 XM_011525882.2 XM_017025614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6855490 c.1229C>A p.P410Q Chr.18: 57550773-57550773 on Build GRCh38
4072700 c.1162G>A p.A388T Chr.18: 57550840-57550840 on Build GRCh38
4623779 c.1246G>A p.V416I Chr.18: 57550756-57550756 on Build GRCh38
7368112 c.1241A>G p.N414S Chr.18: 57550761-57550761 on Build GRCh38
5929819 c.1228C>T p.P410S Chr.18: 57550774-57550774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982999753 Chr.18: 57550657-57550657 on Build GRCh38
rs763703344 Chr.18: 57550689-57550689 on Build GRCh38
rs575034864 Chr.18: 57550668-57550668 on Build GRCh38
rs905014013 Chr.18: 57550654-57550654 on Build GRCh38
rs757452450 Chr.18: 57550774-57550774 on Build GRCh38
rs369783027 Chr.18: 57550808-57550808 on Build GRCh38
rs769121217 Chr.18: 57550714-57550714 on Build GRCh38
rs759682957 Chr.18: 57550863-57550863 on Build GRCh38
rs370894574 Chr.18: 57550661-57550661 on Build GRCh38
rs202192675 Chr.18: 57550712-57550712 on Build GRCh38
rs775784506 Chr.18: 57550665-57550665 on Build GRCh38
rs756233332 Chr.18: 57550829-57550829 on Build GRCh38
rs777848707 Chr.18: 57550704-57550704 on Build GRCh38
rs748481923 Chr.18: 57550850-57550850 on Build GRCh38
rs952591544 Chr.18: 57550652-57550652 on Build GRCh38
rs771713777 Chr.18: 57550855-57550855 on Build GRCh38
rs1626793 Chr.18: 57550873-57550873 on Build GRCh38
rs147782079 Chr.18: 57550832-57550832 on Build GRCh38
rs763340712 Chr.18: 57550688-57550688 on Build GRCh38
rs760250060 Chr.18: 57550859-57550859 on Build GRCh38
rs118204039 Chr.18: 57550734-57550734 on Build GRCh38
rs886053996 Chr.18: 57550652-57550652 on Build GRCh38
rs780160497 Chr.18: 57550697-57550697 on Build GRCh38
rs772796231 Chr.18: 57550733-57550733 on Build GRCh38
rs267606804 Chr.18: 57550759-57550759 on Build GRCh38
rs760290833 Chr.18: 57550743-57550743 on Build GRCh38
rs1625069 Chr.18: 57550874-57550874 on Build GRCh38
rs776285371 Chr.18: 57550872-57550872 on Build GRCh38
rs557575302 Chr.18: 57550677-57550678 on Build GRCh38
rs141118057 Chr.18: 57550836-57550836 on Build GRCh38
rs564058031 Chr.18: 57550706-57550706 on Build GRCh38
rs767086949 Chr.18: 57550731-57550731 on Build GRCh38
rs368654879 Chr.18: 57550694-57550694 on Build GRCh38
rs954352386 Chr.18: 57550792-57550792 on Build GRCh38
rs367564256 Chr.18: 57550763-57550763 on Build GRCh38
rs761457579 Chr.18: 57550729-57550729 on Build GRCh38
rs770247369 Chr.18: 57550711-57550711 on Build GRCh38
rs766054949 Chr.18: 57550868-57550868 on Build GRCh38
rs879482816 Chr.18: 57550873-57550874 on Build GRCh38
rs180762448 Chr.18: 57550703-57550703 on Build GRCh38
rs374230862 Chr.18: 57550693-57550693 on Build GRCh38
rs752448688 Chr.18: 57550773-57550773 on Build GRCh38
rs374298888 Chr.18: 57550690-57550690 on Build GRCh38
rs781276142 Chr.18: 57550709-57550709 on Build GRCh38
rs781302666 Chr.18: 57550785-57550785 on Build GRCh38
rs267606803 Chr.18: 57550760-57550760 on Build GRCh38
rs886053997 Chr.18: 57550657-57550657 on Build GRCh38
rs765020215 Chr.18: 57550772-57550772 on Build GRCh38
rs371363582 Chr.18: 57550771-57550771 on Build GRCh38
rs374760629 Chr.18: 57550707-57550707 on Build GRCh38
rs769047929 Chr.18: 57550837-57550837 on Build GRCh38
rs931949581 Chr.18: 57550727-57550727 on Build GRCh38
rs531300156 Chr.18: 57550724-57550724 on Build GRCh38
rs745970721 Chr.18: 57550710-57550710 on Build GRCh38
rs146899669 Chr.18: 57550753-57550753 on Build GRCh38
rs146918011 Chr.18: 57550838-57550838 on Build GRCh38
rs527375029 Chr.18: 57550700-57550700 on Build GRCh38

View Full Product Details