Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68970138-68970411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX21
Entrez Gene ID
9188
Gene Name
DEAD-box helicase 21
Gene Aliases
GUA, GURDB, RH-II/GU, RH-II/GuA
UniGene
Hs.223141
Gene Chromosome Location
Chr.10: 68956123-68985069 on Build GRCh38
Transcripts
NM_001256910.1 NM_004728.3 XM_011540336.1 XM_017016910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1348759 c.1255C>A p.H419N Chr.10: 68970219-68970219 on Build GRCh38
919845 c.1319G>A p.R440H Chr.10: 68970283-68970283 on Build GRCh38
7171902 c.1312C>G p.Q438E Chr.10: 68970276-68970276 on Build GRCh38
7259181 c.1237-3A>G p.? Chr.10: 68970198-68970198 on Build GRCh38
1203225 c.1318C>T p.R440C Chr.10: 68970282-68970282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768501258 Chr.10: 68970203-68970203 on Build GRCh38
rs777760938 Chr.10: 68970180-68970180 on Build GRCh38
rs779862870 Chr.10: 68970352-68970352 on Build GRCh38
rs766854373 Chr.10: 68970282-68970282 on Build GRCh38
rs371063705 Chr.10: 68970278-68970278 on Build GRCh38
rs768598675 Chr.10: 68970358-68970358 on Build GRCh38
rs150071642 Chr.10: 68970193-68970193 on Build GRCh38
rs376014630 Chr.10: 68970208-68970208 on Build GRCh38
rs761511827 Chr.10: 68970259-68970259 on Build GRCh38
rs753036410 Chr.10: 68970307-68970307 on Build GRCh38
rs34972677 Chr.10: 68970170-68970171 on Build GRCh38
rs781185308 Chr.10: 68970359-68970359 on Build GRCh38
rs987069002 Chr.10: 68970164-68970164 on Build GRCh38
rs756207220 Chr.10: 68970177-68970177 on Build GRCh38
rs749352962 Chr.10: 68970187-68970187 on Build GRCh38
rs201665932 Chr.10: 68970195-68970195 on Build GRCh38
rs778048400 Chr.10: 68970319-68970319 on Build GRCh38
rs769466446 Chr.10: 68970263-68970263 on Build GRCh38
rs747921653 Chr.10: 68970373-68970373 on Build GRCh38
rs777704595 Chr.10: 68970352-68970352 on Build GRCh38
rs150967387 Chr.10: 68970264-68970264 on Build GRCh38
rs201590875 Chr.10: 68970283-68970283 on Build GRCh38
rs759923784 Chr.10: 68970288-68970288 on Build GRCh38
rs769399566 Chr.10: 68970377-68970377 on Build GRCh38
rs767819689 Chr.10: 68970290-68970290 on Build GRCh38
rs769364253 Chr.10: 68970384-68970384 on Build GRCh38
rs746816532 Chr.10: 68970196-68970196 on Build GRCh38
rs771968532 Chr.10: 68970192-68970192 on Build GRCh38
rs763668496 Chr.10: 68970364-68970365 on Build GRCh38
rs762381743 Chr.10: 68970360-68970361 on Build GRCh38
rs746906148 Chr.10: 68970355-68970355 on Build GRCh38
rs764623127 Chr.10: 68970173-68970176 on Build GRCh38
rs373152006 Chr.10: 68970339-68970339 on Build GRCh38
rs772907154 Chr.10: 68970385-68970385 on Build GRCh38
rs757301001 Chr.10: 68970342-68970342 on Build GRCh38
rs756364519 Chr.10: 68970313-68970313 on Build GRCh38

View Full Product Details