Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 157363074-157363347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB6
Entrez Gene ID
10049
Gene Name
DnaJ heat shock protein family (Hsp40) member B6
Gene Aliases
DJ4, DnaJ, HHDJ1, HSJ-2, HSJ2, LGMD1D, LGMD1E, MRJ, MSJ-1
UniGene
Hs.490745
Gene Chromosome Location
Chr.7: 157337004-157417439 on Build GRCh38
Transcripts
NM_005494.2 NM_058246.3 XM_005249515.2 XM_005249516.2 XM_006715823.1 XM_011515704.1 XM_017011633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4980914 c.81A>G p.A27A Chr.7: 157363176-157363176 on Build GRCh38
4980915 c.81A>G p.A27A Chr.7: 157363176-157363176 on Build GRCh38
6324578 c.167T>C p.L56P Chr.7: 157363262-157363262 on Build GRCh38
1087927 c.66-1G>T p.? Chr.7: 157363160-157363160 on Build GRCh38
6324579 c.167T>C p.L56P Chr.7: 157363262-157363262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201730743 Chr.7: 157363217-157363217 on Build GRCh38
rs201246553 Chr.7: 157363305-157363305 on Build GRCh38
rs917989361 Chr.7: 157363324-157363324 on Build GRCh38
rs763891871 Chr.7: 157363314-157363314 on Build GRCh38
rs144525030 Chr.7: 157363176-157363176 on Build GRCh38
rs541772293 Chr.7: 157363312-157363312 on Build GRCh38
rs767485256 Chr.7: 157363298-157363298 on Build GRCh38
rs763596817 Chr.7: 157363209-157363209 on Build GRCh38
rs762681037 Chr.7: 157363191-157363191 on Build GRCh38
rs1007983603 Chr.7: 157363211-157363211 on Build GRCh38
rs948219248 Chr.7: 157363118-157363118 on Build GRCh38
rs200897286 Chr.7: 157363313-157363313 on Build GRCh38
rs371791420 Chr.7: 157363152-157363152 on Build GRCh38
rs775626853 Chr.7: 157363288-157363288 on Build GRCh38
rs140662370 Chr.7: 157363161-157363161 on Build GRCh38
rs575604496 Chr.7: 157363244-157363244 on Build GRCh38
rs779839138 Chr.7: 157363240-157363240 on Build GRCh38
rs770003519 Chr.7: 157363168-157363168 on Build GRCh38
rs747585188 Chr.7: 157363155-157363155 on Build GRCh38
rs533773492 Chr.7: 157363321-157363321 on Build GRCh38
rs148133541 Chr.7: 157363136-157363136 on Build GRCh38
rs200408227 Chr.7: 157363297-157363297 on Build GRCh38
rs141947451 Chr.7: 157363245-157363245 on Build GRCh38
rs776689292 Chr.7: 157363211-157363213 on Build GRCh38
rs771439631 Chr.7: 157363266-157363266 on Build GRCh38
rs772543373 Chr.7: 157363145-157363145 on Build GRCh38
rs370420823 Chr.7: 157363131-157363131 on Build GRCh38
rs751948856 Chr.7: 157363315-157363315 on Build GRCh38
rs1041153586 Chr.7: 157363229-157363229 on Build GRCh38
rs755801484 Chr.7: 157363134-157363134 on Build GRCh38
rs755752166 Chr.7: 157363230-157363230 on Build GRCh38
rs746381911 Chr.7: 157363320-157363320 on Build GRCh38
rs201239906 Chr.7: 157363251-157363251 on Build GRCh38
rs747383972 Chr.7: 157363263-157363263 on Build GRCh38
rs751315424 Chr.7: 157363318-157363318 on Build GRCh38
rs202182637 Chr.7: 157363304-157363304 on Build GRCh38
rs781714119 Chr.7: 157363278-157363278 on Build GRCh38
rs780782893 Chr.7: 157363127-157363127 on Build GRCh38
rs749917279 Chr.7: 157363228-157363228 on Build GRCh38
rs994624000 Chr.7: 157363242-157363242 on Build GRCh38
rs750442450 Chr.7: 157363303-157363303 on Build GRCh38
rs368486917 Chr.7: 157363294-157363294 on Build GRCh38
rs111283304 Chr.7: 157363140-157363140 on Build GRCh38
rs763270397 Chr.7: 157363293-157363293 on Build GRCh38
rs763957260 Chr.7: 157363208-157363208 on Build GRCh38
rs759652351 Chr.7: 157363164-157363164 on Build GRCh38
rs147811155 Chr.7: 157363260-157363260 on Build GRCh38
rs376421671 Chr.7: 157363283-157363283 on Build GRCh38

View Full Product Details