Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59252762-59253035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
UniGene
Hs.607594
Gene Chromosome Location
Chr.10: 59245933-59363186 on Build GRCh38
Transcripts
NM_001001971.2 NM_001143773.1 NM_001166698.1 NM_198215.3 XM_005269618.4 XM_005269619.4 XM_006717702.3 XM_006717703.3 XM_011539490.2 XM_011539491.2 XM_017015885.1 XM_017015886.1 XM_017015887.1 XM_017015888.1 XM_017015889.1 XM_017015890.1 XM_017015891.1 XM_017015892.1 XM_017015893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1702472 c.1523A>G p.E508G Chr.10: 59252808-59252808 on Build GRCh38
5929187 c.1369G>A p.G457R Chr.10: 59252962-59252962 on Build GRCh38
1702473 c.1505C>T p.S502F Chr.10: 59252826-59252826 on Build GRCh38
5689350 c.1363C>A p.P455T Chr.10: 59252968-59252968 on Build GRCh38
3415131 c.1477A>C p.K493Q Chr.10: 59252854-59252854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760701657 Chr.10: 59252884-59252884 on Build GRCh38
rs767997755 Chr.10: 59252936-59252936 on Build GRCh38
rs780471599 Chr.10: 59252796-59252796 on Build GRCh38
rs756157846 Chr.10: 59252940-59252940 on Build GRCh38
rs147340549 Chr.10: 59252845-59252845 on Build GRCh38
rs372356307 Chr.10: 59252801-59252801 on Build GRCh38
rs373089072 Chr.10: 59252928-59252928 on Build GRCh38
rs147832265 Chr.10: 59252971-59252971 on Build GRCh38
rs775100071 Chr.10: 59252881-59252881 on Build GRCh38
rs193920897 Chr.10: 59252904-59252904 on Build GRCh38
rs147212027 Chr.10: 59252836-59252836 on Build GRCh38
rs1002338139 Chr.10: 59252887-59252887 on Build GRCh38
rs144395945 Chr.10: 59252900-59252900 on Build GRCh38
rs746726052 Chr.10: 59253000-59253000 on Build GRCh38
rs752842174 Chr.10: 59252938-59252938 on Build GRCh38
rs765018746 Chr.10: 59252906-59252906 on Build GRCh38
rs772896641 Chr.10: 59252909-59252909 on Build GRCh38
rs766794321 Chr.10: 59252921-59252921 on Build GRCh38
rs923034389 Chr.10: 59252829-59252829 on Build GRCh38
rs940280885 Chr.10: 59252865-59252865 on Build GRCh38
rs779907338 Chr.10: 59252987-59252987 on Build GRCh38
rs762459660 Chr.10: 59252913-59252913 on Build GRCh38
rs755489345 Chr.10: 59252931-59252931 on Build GRCh38
rs369226393 Chr.10: 59252970-59252970 on Build GRCh38
rs867381443 Chr.10: 59252980-59252980 on Build GRCh38
rs752126123 Chr.10: 59252799-59252799 on Build GRCh38
rs867251521 Chr.10: 59252967-59252967 on Build GRCh38
rs938184648 Chr.10: 59252950-59252950 on Build GRCh38
rs768651858 Chr.10: 59252886-59252886 on Build GRCh38
rs933080663 Chr.10: 59252832-59252832 on Build GRCh38
rs376088168 Chr.10: 59252966-59252966 on Build GRCh38
rs781254036 Chr.10: 59252824-59252824 on Build GRCh38
rs914203068 Chr.10: 59252871-59252871 on Build GRCh38
rs199877009 Chr.10: 59252893-59252893 on Build GRCh38
rs141046953 Chr.10: 59252878-59252878 on Build GRCh38

View Full Product Details