Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 140448660-140448885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN2
Entrez Gene ID
64084
Gene Name
calsyntenin 2
Gene Aliases
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma
UniGene
Hs.158529
Gene Chromosome Location
Chr.3: 139935185-140577397 on Build GRCh38
Transcripts
NM_022131.2 XM_017007022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
728127 c.929G>C p.C310S Chr.3: 140448660-140448660 on Build GRCh38
3915034 c.931G>A p.D311N Chr.3: 140448662-140448662 on Build GRCh38
4825492 c.958C>G p.L320V Chr.3: 140448689-140448689 on Build GRCh38
3124419 c.935G>A p.R312Q Chr.3: 140448666-140448666 on Build GRCh38
3588114 c.936G>A p.R312R Chr.3: 140448667-140448667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550139289 Chr.3: 140448807-140448807 on Build GRCh38
rs886965769 Chr.3: 140448761-140448761 on Build GRCh38
rs763973636 Chr.3: 140448721-140448721 on Build GRCh38
rs745392675 Chr.3: 140448757-140448757 on Build GRCh38
rs1036896197 Chr.3: 140448683-140448683 on Build GRCh38
rs760597356 Chr.3: 140448717-140448717 on Build GRCh38
rs898231885 Chr.3: 140448688-140448688 on Build GRCh38
rs1031875900 Chr.3: 140448805-140448805 on Build GRCh38
rs1039583111 Chr.3: 140448710-140448710 on Build GRCh38
rs757221407 Chr.3: 140448725-140448725 on Build GRCh38
rs999419022 Chr.3: 140448802-140448802 on Build GRCh38
rs899765554 Chr.3: 140448714-140448714 on Build GRCh38
rs532023239 Chr.3: 140448779-140448779 on Build GRCh38
rs1033914338 Chr.3: 140448843-140448843 on Build GRCh38
rs750559755 Chr.3: 140448741-140448741 on Build GRCh38
rs753538234 Chr.3: 140448724-140448724 on Build GRCh38
rs568645028 Chr.3: 140448828-140448828 on Build GRCh38
rs566620886 Chr.3: 140448852-140448852 on Build GRCh38
rs369596942 Chr.3: 140448710-140448710 on Build GRCh38
rs752366536 Chr.3: 140448708-140448708 on Build GRCh38
rs759107918 Chr.3: 140448687-140448687 on Build GRCh38
rs564612941 Chr.3: 140448769-140448769 on Build GRCh38
rs773928562 Chr.3: 140448686-140448686 on Build GRCh38
rs529705913 Chr.3: 140448842-140448842 on Build GRCh38
rs907478049 Chr.3: 140448771-140448771 on Build GRCh38
rs1028298538 Chr.3: 140448861-140448861 on Build GRCh38
rs957401203 Chr.3: 140448814-140448814 on Build GRCh38
rs2081803 Chr.3: 140448694-140448694 on Build GRCh38
rs374267620 Chr.3: 140448754-140448754 on Build GRCh38
rs144791859 Chr.3: 140448795-140448795 on Build GRCh38
rs533740737 Chr.3: 140448855-140448855 on Build GRCh38
rs78744687 Chr.3: 140448851-140448851 on Build GRCh38
rs778657160 Chr.3: 140448735-140448735 on Build GRCh38

View Full Product Details