Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74098542-74098799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL2
Entrez Gene ID
2920
Gene Name
C-X-C motif chemokine ligand 2
Gene Aliases
CINC-2a, GRO2, GROb, MGSA-b, MIP-2a, MIP2, MIP2A, SCYB2
UniGene
Hs.75765
Gene Chromosome Location
Chr.4: 74097035-74099280 on Build GRCh38
Transcripts
NM_002089.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1057484 c.292G>A p.E98K Chr.4: 74098617-74098617 on Build GRCh38
1057485 c.225-6C>A p.? Chr.4: 74098690-74098690 on Build GRCh38
4125920 c.232C>T p.L78F Chr.4: 74098677-74098677 on Build GRCh38
7094456 c.291C>T p.I97I Chr.4: 74098618-74098618 on Build GRCh38
6279968 c.270G>C p.S90S Chr.4: 74098639-74098639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749310499 Chr.4: 74098702-74098702 on Build GRCh38
rs771936739 Chr.4: 74098603-74098603 on Build GRCh38
rs6832638 Chr.4: 74098611-74098611 on Build GRCh38
rs930350156 Chr.4: 74098674-74098674 on Build GRCh38
rs758110362 Chr.4: 74098654-74098654 on Build GRCh38
rs371945474 Chr.4: 74098762-74098762 on Build GRCh38
rs987337193 Chr.4: 74098563-74098563 on Build GRCh38
rs199752288 Chr.4: 74098721-74098721 on Build GRCh38
rs200665196 Chr.4: 74098777-74098777 on Build GRCh38
rs756460806 Chr.4: 74098667-74098667 on Build GRCh38
rs201534860 Chr.4: 74098729-74098729 on Build GRCh38
rs762044384 Chr.4: 74098638-74098638 on Build GRCh38
rs746075939 Chr.4: 74098660-74098660 on Build GRCh38
rs201352382 Chr.4: 74098711-74098711 on Build GRCh38
rs199529315 Chr.4: 74098768-74098768 on Build GRCh38
rs764500980 Chr.4: 74098698-74098698 on Build GRCh38
rs553596447 Chr.4: 74098773-74098773 on Build GRCh38
rs755085970 Chr.4: 74098779-74098779 on Build GRCh38
rs757118828 Chr.4: 74098764-74098764 on Build GRCh38
rs188944956 Chr.4: 74098726-74098726 on Build GRCh38
rs775781780 Chr.4: 74098609-74098609 on Build GRCh38
rs754490441 Chr.4: 74098646-74098646 on Build GRCh38
rs752706721 Chr.4: 74098648-74098648 on Build GRCh38
rs371227293 Chr.4: 74098706-74098706 on Build GRCh38
rs200873298 Chr.4: 74098591-74098591 on Build GRCh38
rs745343380 Chr.4: 74098601-74098601 on Build GRCh38
rs202141695 Chr.4: 74098693-74098693 on Build GRCh38
rs911702801 Chr.4: 74098587-74098587 on Build GRCh38
rs568060950 Chr.4: 74098749-74098749 on Build GRCh38
rs538428386 Chr.4: 74098645-74098645 on Build GRCh38
rs201623380 Chr.4: 74098690-74098690 on Build GRCh38
rs747835145 Chr.4: 74098600-74098600 on Build GRCh38
rs748738093 Chr.4: 74098704-74098704 on Build GRCh38
rs758998062 Chr.4: 74098727-74098727 on Build GRCh38
rs368385936 Chr.4: 74098753-74098753 on Build GRCh38
rs200320149 Chr.4: 74098688-74098688 on Build GRCh38
rs780434409 Chr.4: 74098696-74098696 on Build GRCh38
rs377707814 Chr.4: 74098644-74098644 on Build GRCh38
rs777667311 Chr.4: 74098659-74098659 on Build GRCh38
rs200214601 Chr.4: 74098703-74098703 on Build GRCh38
rs535488364 Chr.4: 74098759-74098759 on Build GRCh38
rs775002602 Chr.4: 74098635-74098635 on Build GRCh38
rs1003361647 Chr.4: 74098680-74098680 on Build GRCh38
rs778838611 Chr.4: 74098596-74098596 on Build GRCh38
rs146575633 Chr.4: 74098647-74098647 on Build GRCh38
rs760413015 Chr.4: 74098640-74098640 on Build GRCh38
rs769965449 Chr.4: 74098607-74098607 on Build GRCh38
rs1045085792 Chr.4: 74098677-74098677 on Build GRCh38
rs758430569 Chr.4: 74098757-74098757 on Build GRCh38
rs201836008 Chr.4: 74098604-74098604 on Build GRCh38
rs769226365 Chr.4: 74098624-74098624 on Build GRCh38
rs200989587 Chr.4: 74098639-74098639 on Build GRCh38
rs760962805 Chr.4: 74098713-74098713 on Build GRCh38

View Full Product Details