Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112827106-112827361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD200R1L
Entrez Gene ID
344807
Gene Name
CD200 receptor 1 like
Gene Aliases
CD200R2, CD200RLa
UniGene
Hs.531814
Gene Chromosome Location
Chr.3: 112814925-112847004 on Build GRCh38
Transcripts
NM_001008784.2 NM_001199215.1 XM_011512763.2 XM_017006300.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4760203 c.521T>A p.I174N Chr.3: 112827151-112827151 on Build GRCh38
6885475 c.487G>A p.V163I Chr.3: 112827185-112827185 on Build GRCh38
255899 c.484G>C p.A162P Chr.3: 112827188-112827188 on Build GRCh38
1200263 c.538C>A p.L180I Chr.3: 112827134-112827134 on Build GRCh38
3585417 c.526G>T p.E176* Chr.3: 112827146-112827146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767794455 Chr.3: 112827318-112827318 on Build GRCh38
rs781599292 Chr.3: 112827146-112827146 on Build GRCh38
rs764454316 Chr.3: 112827188-112827188 on Build GRCh38
rs768130659 Chr.3: 112827252-112827252 on Build GRCh38
rs781616615 Chr.3: 112827214-112827214 on Build GRCh38
rs1009276419 Chr.3: 112827134-112827134 on Build GRCh38
rs987132191 Chr.3: 112827182-112827182 on Build GRCh38
rs766303593 Chr.3: 112827280-112827280 on Build GRCh38
rs750760135 Chr.3: 112827319-112827319 on Build GRCh38
rs867375041 Chr.3: 112827218-112827218 on Build GRCh38
rs779042191 Chr.3: 112827130-112827130 on Build GRCh38
rs764258316 Chr.3: 112827288-112827288 on Build GRCh38
rs991316634 Chr.3: 112827335-112827335 on Build GRCh38
rs774046786 Chr.3: 112827259-112827259 on Build GRCh38
rs779461488 Chr.3: 112827234-112827234 on Build GRCh38
rs146131812 Chr.3: 112827144-112827144 on Build GRCh38
rs778306942 Chr.3: 112827137-112827137 on Build GRCh38
rs572324262 Chr.3: 112827260-112827261 on Build GRCh38
rs200707844 Chr.3: 112827274-112827274 on Build GRCh38
rs774148636 Chr.3: 112827246-112827247 on Build GRCh38
rs769254542 Chr.3: 112827151-112827151 on Build GRCh38
rs772998031 Chr.3: 112827273-112827273 on Build GRCh38
rs371039716 Chr.3: 112827169-112827169 on Build GRCh38
rs773959702 Chr.3: 112827154-112827154 on Build GRCh38
rs767283846 Chr.3: 112827166-112827166 on Build GRCh38
rs531343747 Chr.3: 112827132-112827132 on Build GRCh38
rs369357384 Chr.3: 112827138-112827138 on Build GRCh38
rs566932430 Chr.3: 112827261-112827261 on Build GRCh38
rs746373428 Chr.3: 112827148-112827148 on Build GRCh38
rs547208713 Chr.3: 112827202-112827202 on Build GRCh38
rs367755232 Chr.3: 112827177-112827177 on Build GRCh38
rs775310729 Chr.3: 112827261-112827261 on Build GRCh38
rs371433258 Chr.3: 112827316-112827316 on Build GRCh38
rs751810985 Chr.3: 112827195-112827195 on Build GRCh38
rs140173974 Chr.3: 112827173-112827173 on Build GRCh38
rs761642930 Chr.3: 112827162-112827162 on Build GRCh38
rs776404385 Chr.3: 112827283-112827283 on Build GRCh38
rs756594504 Chr.3: 112827233-112827233 on Build GRCh38
rs747791809 Chr.3: 112827260-112827260 on Build GRCh38
rs762535035 Chr.3: 112827153-112827153 on Build GRCh38
rs958558995 Chr.3: 112827308-112827308 on Build GRCh38
rs760422252 Chr.3: 112827268-112827268 on Build GRCh38
rs759454830 Chr.3: 112827258-112827258 on Build GRCh38
rs746366169 Chr.3: 112827229-112827229 on Build GRCh38
rs540129972 Chr.3: 112827186-112827186 on Build GRCh38
rs758638949 Chr.3: 112827133-112827133 on Build GRCh38
rs775007452 Chr.3: 112827152-112827152 on Build GRCh38
rs536022172 Chr.3: 112827290-112827290 on Build GRCh38

View Full Product Details