Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134244661-134244934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC160
Entrez Gene ID
347475
Gene Name
coiled-coil domain containing 160
Gene Aliases
-
UniGene
Hs.679431
Gene Chromosome Location
Chr.X: 134237047-134246207 on Build GRCh38
Transcripts
NM_001101357.1 XM_005262409.4 XM_005262410.4

SNP Detail

SNP ID Chromosome Location
rs753404055 Chr.X: 134244836-134244836 on Build GRCh38
rs756936691 Chr.X: 134244752-134244752 on Build GRCh38
rs1052155702 Chr.X: 134244744-134244744 on Build GRCh38
rs752374882 Chr.X: 134244760-134244760 on Build GRCh38
rs201693622 Chr.X: 134244892-134244892 on Build GRCh38
rs11798214 Chr.X: 134244880-134244880 on Build GRCh38
rs749574186 Chr.X: 134244855-134244855 on Build GRCh38
rs746701213 Chr.X: 134244690-134244690 on Build GRCh38
rs780482621 Chr.X: 134244848-134244848 on Build GRCh38
rs925440373 Chr.X: 134244860-134244860 on Build GRCh38
rs925169605 Chr.X: 134244691-134244691 on Build GRCh38
rs748338127 Chr.X: 134244878-134244878 on Build GRCh38
rs763616585 Chr.X: 134244791-134244791 on Build GRCh38
rs759856987 Chr.X: 134244891-134244893 on Build GRCh38
rs947108162 Chr.X: 134244814-134244814 on Build GRCh38
rs759528450 Chr.X: 134244908-134244908 on Build GRCh38
rs375551602 Chr.X: 134244907-134244907 on Build GRCh38
rs6638192 Chr.X: 134244808-134244808 on Build GRCh38
rs936481504 Chr.X: 134244739-134244739 on Build GRCh38
rs756880198 Chr.X: 134244802-134244802 on Build GRCh38
rs200879811 Chr.X: 134244862-134244862 on Build GRCh38
rs762673363 Chr.X: 134244773-134244773 on Build GRCh38
rs772201502 Chr.X: 134244891-134244891 on Build GRCh38
rs1040752146 Chr.X: 134244890-134244890 on Build GRCh38
rs780818748 Chr.X: 134244820-134244820 on Build GRCh38
rs751140112 Chr.X: 134244798-134244798 on Build GRCh38
rs368396449 Chr.X: 134244778-134244778 on Build GRCh38
rs779154022 Chr.X: 134244869-134244869 on Build GRCh38
rs370691264 Chr.X: 134244867-134244867 on Build GRCh38
rs756568036 Chr.X: 134244828-134244828 on Build GRCh38
rs1007857969 Chr.X: 134244864-134244864 on Build GRCh38

View Full Product Details