Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34106339-34106596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF12
Entrez Gene ID
25853
Gene Name
DDB1 and CUL4 associated factor 12
Gene Aliases
CT102, KIAA1892, TCC52, WDR40A
UniGene
Hs.493750
Gene Chromosome Location
Chr.9: 34086383-34126773 on Build GRCh38
Transcripts
NM_015397.3 XM_011517843.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640345 c.583G>A p.D195N Chr.9: 34106452-34106452 on Build GRCh38
6640346 c.529G>A p.D177N Chr.9: 34106452-34106452 on Build GRCh38
4373896 c.517G>A p.A173T Chr.9: 34106464-34106464 on Build GRCh38
3395796 c.546A>G p.G182G Chr.9: 34106489-34106489 on Build GRCh38
3395797 c.492A>G p.G164G Chr.9: 34106489-34106489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376139003 Chr.9: 34106464-34106464 on Build GRCh38
rs765203748 Chr.9: 34106387-34106387 on Build GRCh38
rs769970332 Chr.9: 34106500-34106500 on Build GRCh38
rs147332697 Chr.9: 34106453-34106453 on Build GRCh38
rs151311642 Chr.9: 34106492-34106492 on Build GRCh38
rs747327833 Chr.9: 34106465-34106465 on Build GRCh38
rs550259873 Chr.9: 34106359-34106359 on Build GRCh38
rs370693262 Chr.9: 34106542-34106542 on Build GRCh38
rs1045583210 Chr.9: 34106552-34106552 on Build GRCh38
rs377329450 Chr.9: 34106428-34106428 on Build GRCh38
rs759624402 Chr.9: 34106384-34106384 on Build GRCh38
rs539284048 Chr.9: 34106502-34106502 on Build GRCh38
rs771177118 Chr.9: 34106489-34106489 on Build GRCh38
rs761658919 Chr.9: 34106530-34106530 on Build GRCh38
rs767388718 Chr.9: 34106531-34106531 on Build GRCh38
rs371173684 Chr.9: 34106554-34106554 on Build GRCh38
rs752601145 Chr.9: 34106510-34106510 on Build GRCh38
rs780941540 Chr.9: 34106432-34106432 on Build GRCh38
rs745401504 Chr.9: 34106438-34106438 on Build GRCh38
rs781554437 Chr.9: 34106526-34106526 on Build GRCh38
rs376012211 Chr.9: 34106559-34106559 on Build GRCh38
rs957458019 Chr.9: 34106462-34106462 on Build GRCh38
rs889118203 Chr.9: 34106478-34106478 on Build GRCh38
rs752725051 Chr.9: 34106395-34106395 on Build GRCh38
rs762949620 Chr.9: 34106516-34106516 on Build GRCh38
rs764926887 Chr.9: 34106506-34106506 on Build GRCh38
rs560691606 Chr.9: 34106402-34106402 on Build GRCh38
rs140591380 Chr.9: 34106494-34106494 on Build GRCh38
rs773165467 Chr.9: 34106411-34106411 on Build GRCh38
rs779416665 Chr.9: 34106450-34106450 on Build GRCh38
rs867263881 Chr.9: 34106468-34106469 on Build GRCh38
rs751420114 Chr.9: 34106522-34106522 on Build GRCh38
rs747020535 Chr.9: 34106452-34106452 on Build GRCh38
rs977588723 Chr.9: 34106425-34106425 on Build GRCh38
rs750149911 Chr.9: 34106545-34106545 on Build GRCh38
rs544621358 Chr.9: 34106446-34106446 on Build GRCh38
rs10971917 Chr.9: 34106412-34106412 on Build GRCh38
rs756982062 Chr.9: 34106424-34106424 on Build GRCh38
rs764063868 Chr.9: 34106521-34106521 on Build GRCh38

View Full Product Details