Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 51027850-51028115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COBL
Entrez Gene ID
23242
Gene Name
cordon-bleu WH2 repeat protein
Gene Aliases
-
UniGene
Hs.99141
Gene Chromosome Location
Chr.7: 51016212-51316818 on Build GRCh38
Transcripts
NM_001287436.1 NM_015198.3 XM_005271750.1 XM_005271751.1 XM_005271756.1 XM_011515234.1 XM_011515235.1 XM_011515236.1 XM_011515237.1 XM_011515238.1 XM_011515239.1 XM_011515240.1 XM_011515241.1 XM_017011898.1 XM_017011899.1 XM_017011900.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1090687 c.3230G>A p.G1077E Chr.7: 51027866-51027866 on Build GRCh38
1248699 c.3127G>A p.A1043T Chr.7: 51027969-51027969 on Build GRCh38
301964 c.3125G>A p.R1042H Chr.7: 51027971-51027971 on Build GRCh38
4626045 c.3214G>A p.V1072M Chr.7: 51027882-51027882 on Build GRCh38
4905366 c.3487G>A p.D1163N Chr.7: 51027855-51027855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564872553 Chr.7: 51027969-51027969 on Build GRCh38
rs141696883 Chr.7: 51028054-51028054 on Build GRCh38
rs17134127 Chr.7: 51028053-51028053 on Build GRCh38
rs765381841 Chr.7: 51027882-51027882 on Build GRCh38
rs756926554 Chr.7: 51027990-51027990 on Build GRCh38
rs765992520 Chr.7: 51028080-51028080 on Build GRCh38
rs895793227 Chr.7: 51028068-51028068 on Build GRCh38
rs150541791 Chr.7: 51028056-51028056 on Build GRCh38
rs770049726 Chr.7: 51028023-51028023 on Build GRCh38
rs200478913 Chr.7: 51028055-51028055 on Build GRCh38
rs191244742 Chr.7: 51027955-51027955 on Build GRCh38
rs780099196 Chr.7: 51028001-51028001 on Build GRCh38
rs199919808 Chr.7: 51027962-51027962 on Build GRCh38
rs766864509 Chr.7: 51027897-51027897 on Build GRCh38
rs374579503 Chr.7: 51027912-51027912 on Build GRCh38
rs764853569 Chr.7: 51027994-51027994 on Build GRCh38
rs1026269111 Chr.7: 51027895-51027895 on Build GRCh38
rs749362981 Chr.7: 51028040-51028040 on Build GRCh38
rs201041607 Chr.7: 51027938-51027938 on Build GRCh38
rs574131063 Chr.7: 51027885-51027885 on Build GRCh38
rs376515872 Chr.7: 51028066-51028066 on Build GRCh38
rs747950142 Chr.7: 51028022-51028022 on Build GRCh38
rs745433161 Chr.7: 51027947-51027947 on Build GRCh38
rs776944424 Chr.7: 51027960-51027960 on Build GRCh38
rs781139732 Chr.7: 51028011-51028011 on Build GRCh38
rs910301712 Chr.7: 51028047-51028047 on Build GRCh38
rs749925541 Chr.7: 51027997-51027997 on Build GRCh38
rs934903243 Chr.7: 51027887-51027887 on Build GRCh38
rs201043019 Chr.7: 51027970-51027970 on Build GRCh38
rs117542691 Chr.7: 51027891-51027891 on Build GRCh38
rs547420267 Chr.7: 51027978-51027978 on Build GRCh38
rs746769073 Chr.7: 51027957-51027957 on Build GRCh38
rs144463257 Chr.7: 51027999-51027999 on Build GRCh38
rs773343530 Chr.7: 51028029-51028029 on Build GRCh38
rs999908607 Chr.7: 51028018-51028018 on Build GRCh38
rs370605120 Chr.7: 51028069-51028069 on Build GRCh38
rs373944191 Chr.7: 51028086-51028086 on Build GRCh38
rs772030687 Chr.7: 51027954-51027954 on Build GRCh38
rs138933352 Chr.7: 51027961-51027961 on Build GRCh38
rs968465295 Chr.7: 51028008-51028008 on Build GRCh38
rs753525093 Chr.7: 51027936-51027936 on Build GRCh38
rs948771797 Chr.7: 51027939-51027939 on Build GRCh38
rs201511321 Chr.7: 51027982-51027982 on Build GRCh38
rs746965445 Chr.7: 51028004-51028004 on Build GRCh38
rs751483466 Chr.7: 51028082-51028082 on Build GRCh38
rs779159023 Chr.7: 51027940-51027940 on Build GRCh38
rs956164036 Chr.7: 51028037-51028037 on Build GRCh38
rs370348190 Chr.7: 51027972-51027972 on Build GRCh38
rs768602767 Chr.7: 51027958-51027958 on Build GRCh38
rs901070611 Chr.7: 51027983-51027983 on Build GRCh38
rs750067179 Chr.7: 51028067-51028067 on Build GRCh38
rs142891237 Chr.7: 51027968-51027968 on Build GRCh38
rs777376866 Chr.7: 51027916-51027916 on Build GRCh38

View Full Product Details