Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59181671-59181942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX4
Entrez Gene ID
23220
Gene Name
deltex E3 ubiquitin ligase 4
Gene Aliases
RNF155
UniGene
Hs.523696
Gene Chromosome Location
Chr.11: 59171430-59208587 on Build GRCh38
Transcripts
NM_001300727.1 NM_015177.1 XM_006718483.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4679624 c.233G>A p.R78H Chr.11: 59181760-59181760 on Build GRCh38
5530251 c.93C>T p.F31F Chr.11: 59181938-59181938 on Build GRCh38
4679625 c.332T>C p.I111T Chr.11: 59181859-59181859 on Build GRCh38
6650104 c.316G>A p.D106N Chr.11: 59181843-59181843 on Build GRCh38
5889142 c.278G>A p.W93* Chr.11: 59181805-59181805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576672770 Chr.11: 59181708-59181708 on Build GRCh38
rs199904268 Chr.11: 59181725-59181725 on Build GRCh38
rs778910305 Chr.11: 59181862-59181862 on Build GRCh38
rs761374600 Chr.11: 59181748-59181748 on Build GRCh38
rs746807888 Chr.11: 59181829-59181829 on Build GRCh38
rs771899968 Chr.11: 59181727-59181727 on Build GRCh38
rs755502863 Chr.11: 59181759-59181759 on Build GRCh38
rs373943632 Chr.11: 59181784-59181784 on Build GRCh38
rs756322208 Chr.11: 59181781-59181781 on Build GRCh38
rs753046961 Chr.11: 59181697-59181697 on Build GRCh38
rs781563130 Chr.11: 59181760-59181760 on Build GRCh38
rs760038957 Chr.11: 59181838-59181838 on Build GRCh38
rs777782871 Chr.11: 59181705-59181705 on Build GRCh38
rs747017849 Chr.11: 59181894-59181894 on Build GRCh38
rs534485061 Chr.11: 59181913-59181913 on Build GRCh38
rs762336958 Chr.11: 59181912-59181912 on Build GRCh38
rs779035023 Chr.11: 59181794-59181794 on Build GRCh38
rs202222554 Chr.11: 59181733-59181733 on Build GRCh38
rs753285278 Chr.11: 59181843-59181843 on Build GRCh38
rs762732859 Chr.11: 59181730-59181730 on Build GRCh38
rs779827794 Chr.11: 59181892-59181892 on Build GRCh38
rs950608700 Chr.11: 59181803-59181803 on Build GRCh38
rs771913438 Chr.11: 59181800-59181800 on Build GRCh38
rs745756061 Chr.11: 59181798-59181798 on Build GRCh38
rs776635786 Chr.11: 59181899-59181899 on Build GRCh38
rs764652936 Chr.11: 59181746-59181746 on Build GRCh38
rs376164008 Chr.11: 59181842-59181842 on Build GRCh38
rs377422945 Chr.11: 59181720-59181720 on Build GRCh38
rs752146380 Chr.11: 59181792-59181792 on Build GRCh38
rs573430937 Chr.11: 59181856-59181856 on Build GRCh38
rs370352867 Chr.11: 59181728-59181728 on Build GRCh38
rs539999797 Chr.11: 59181835-59181835 on Build GRCh38
rs373681418 Chr.11: 59181818-59181818 on Build GRCh38
rs758319344 Chr.11: 59181878-59181878 on Build GRCh38
rs769592534 Chr.11: 59181907-59181907 on Build GRCh38
rs571644429 Chr.11: 59181744-59181744 on Build GRCh38
rs747737410 Chr.11: 59181900-59181900 on Build GRCh38
rs745960840 Chr.11: 59181872-59181872 on Build GRCh38
rs764887811 Chr.11: 59181756-59181756 on Build GRCh38
rs555328446 Chr.11: 59181707-59181707 on Build GRCh38
rs756410632 Chr.11: 59181855-59181855 on Build GRCh38
rs772737375 Chr.11: 59181909-59181909 on Build GRCh38
rs370390470 Chr.11: 59181770-59181770 on Build GRCh38
rs368146768 Chr.11: 59181758-59181758 on Build GRCh38
rs753027022 Chr.11: 59181775-59181775 on Build GRCh38
rs978082454 Chr.11: 59181797-59181797 on Build GRCh38
rs558443267 Chr.11: 59181836-59181836 on Build GRCh38
rs992629909 Chr.11: 59181769-59181769 on Build GRCh38
rs370029050 Chr.11: 59181734-59181734 on Build GRCh38
rs768609170 Chr.11: 59181831-59181831 on Build GRCh38
rs754051185 Chr.11: 59181857-59181857 on Build GRCh38
rs752098525 Chr.11: 59181757-59181757 on Build GRCh38
rs768559763 Chr.11: 59181895-59181895 on Build GRCh38
rs368390838 Chr.11: 59181787-59181787 on Build GRCh38
rs753356918 Chr.11: 59181747-59181747 on Build GRCh38
rs759531436 Chr.11: 59181726-59181726 on Build GRCh38

View Full Product Details