Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124986597-124986856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC15
Entrez Gene ID
80071
Gene Name
coiled-coil domain containing 15
Gene Aliases
-
UniGene
Hs.287555
Gene Chromosome Location
Chr.11: 124954121-125041489 on Build GRCh38
Transcripts
NM_025004.2 XM_017018346.1 XM_017018347.1 XM_017018348.1 XM_017018349.1 XM_017018350.1 XM_017018351.1 XM_017018352.1 XM_017018353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
924483 c.794C>A p.S265Y Chr.11: 124986782-124986782 on Build GRCh38
4778407 c.863A>G p.H288R Chr.11: 124986851-124986851 on Build GRCh38
5696862 c.805G>A p.D269N Chr.11: 124986793-124986793 on Build GRCh38
6880484 c.768G>A p.E256E Chr.11: 124986756-124986756 on Build GRCh38
1737638 c.847G>A p.D283N Chr.11: 124986835-124986835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757150643 Chr.11: 124986709-124986710 on Build GRCh38
rs776810547 Chr.11: 124986747-124986761 on Build GRCh38
rs749747835 Chr.11: 124986725-124986725 on Build GRCh38
rs897741320 Chr.11: 124986799-124986799 on Build GRCh38
rs543296409 Chr.11: 124986718-124986718 on Build GRCh38
rs563956294 Chr.11: 124986771-124986771 on Build GRCh38
rs371629051 Chr.11: 124986704-124986704 on Build GRCh38
rs772127489 Chr.11: 124986715-124986716 on Build GRCh38
rs753690576 Chr.11: 124986711-124986711 on Build GRCh38
rs879156271 Chr.11: 124986720-124986720 on Build GRCh38
rs983926243 Chr.11: 124986684-124986684 on Build GRCh38
rs974864604 Chr.11: 124986701-124986702 on Build GRCh38
rs370378819 Chr.11: 124986689-124986689 on Build GRCh38
rs904738184 Chr.11: 124986760-124986760 on Build GRCh38
rs373335617 Chr.11: 124986806-124986806 on Build GRCh38
rs753459165 Chr.11: 124986686-124986688 on Build GRCh38
rs114780933 Chr.11: 124986709-124986709 on Build GRCh38
rs767217050 Chr.11: 124986704-124986705 on Build GRCh38
rs748417722 Chr.11: 124986717-124986717 on Build GRCh38
rs752274998 Chr.11: 124986705-124986706 on Build GRCh38
rs80062218 Chr.11: 124986756-124986756 on Build GRCh38
rs764033281 Chr.11: 124986710-124986710 on Build GRCh38
rs535328688 Chr.11: 124986690-124986690 on Build GRCh38
rs963418825 Chr.11: 124986694-124986694 on Build GRCh38
rs765318050 Chr.11: 124986688-124986691 on Build GRCh38
rs11219860 Chr.11: 124986691-124986691 on Build GRCh38
rs777233071 Chr.11: 124986698-124986699 on Build GRCh38
rs369194843 Chr.11: 124986707-124986707 on Build GRCh38
rs557905302 Chr.11: 124986706-124986706 on Build GRCh38
rs775850200 Chr.11: 124986716-124986717 on Build GRCh38
rs757453582 Chr.11: 124986703-124986703 on Build GRCh38
rs11219861 Chr.11: 124986693-124986693 on Build GRCh38
rs569534952 Chr.11: 124986688-124986688 on Build GRCh38
rs769112544 Chr.11: 124986729-124986729 on Build GRCh38
rs543576074 Chr.11: 124986708-124986708 on Build GRCh38
rs760446480 Chr.11: 124986826-124986826 on Build GRCh38
rs866328911 Chr.11: 124986700-124986700 on Build GRCh38
rs951393048 Chr.11: 124986682-124986691 on Build GRCh38
rs60614383 Chr.11: 124986719-124986720 on Build GRCh38
rs565690661 Chr.11: 124986698-124986698 on Build GRCh38
rs11219862 Chr.11: 124986702-124986702 on Build GRCh38
rs34449008 Chr.11: 124986690-124986691 on Build GRCh38
rs777410517 Chr.11: 124986708-124986720 on Build GRCh38
rs994141710 Chr.11: 124986701-124986701 on Build GRCh38
rs549924386 Chr.11: 124986650-124986650 on Build GRCh38
rs1055621024 Chr.11: 124986626-124986626 on Build GRCh38
rs370197049 Chr.11: 124986803-124986803 on Build GRCh38
rs994609440 Chr.11: 124986817-124986817 on Build GRCh38
rs745331772 Chr.11: 124986798-124986798 on Build GRCh38
rs758680911 Chr.11: 124986703-124986704 on Build GRCh38
rs746823651 Chr.11: 124986707-124986716 on Build GRCh38
rs771413217 Chr.11: 124986736-124986736 on Build GRCh38
rs766008960 Chr.11: 124986703-124986714 on Build GRCh38
rs757274427 Chr.11: 124986712-124986712 on Build GRCh38
rs760655845 Chr.11: 124986668-124986671 on Build GRCh38
rs781342914 Chr.11: 124986716-124986716 on Build GRCh38
rs773971332 Chr.11: 124986699-124986699 on Build GRCh38
rs749273998 Chr.11: 124986709-124986714 on Build GRCh38
rs146292043 Chr.11: 124986668-124986669 on Build GRCh38
rs895759167 Chr.11: 124986667-124986667 on Build GRCh38
rs768500514 Chr.11: 124986707-124986714 on Build GRCh38
rs1055927815 Chr.11: 124986746-124986746 on Build GRCh38
rs763553668 Chr.11: 124986696-124986696 on Build GRCh38
rs878887902 Chr.11: 124986700-124986701 on Build GRCh38
rs779128005 Chr.11: 124986705-124986714 on Build GRCh38
rs747169607 Chr.11: 124986718-124986719 on Build GRCh38
rs191035729 Chr.11: 124986747-124986747 on Build GRCh38
rs755076801 Chr.11: 124986715-124986715 on Build GRCh38
rs1025584564 Chr.11: 124986655-124986655 on Build GRCh38
rs1036284900 Chr.11: 124986774-124986774 on Build GRCh38
rs776650364 Chr.11: 124986686-124986691 on Build GRCh38
rs374859694 Chr.11: 124986705-124986705 on Build GRCh38
rs35483983 Chr.11: 124986691-124986692 on Build GRCh38
rs745890127 Chr.11: 124986713-124986714 on Build GRCh38
rs373279319 Chr.11: 124986719-124986719 on Build GRCh38
rs374160264 Chr.11: 124986667-124986668 on Build GRCh38
rs376142029 Chr.11: 124986711-124986714 on Build GRCh38
rs567665602 Chr.11: 124986670-124986671 on Build GRCh38

View Full Product Details