Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18899492-18899741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPE
Entrez Gene ID
11316
Gene Name
coatomer protein complex subunit epsilon
Gene Aliases
epsilon-COP
UniGene
Hs.10326
Gene Chromosome Location
Chr.19: 18899514-18919464 on Build GRCh38
Transcripts
NM_007263.3 NM_199442.1 NM_199444.1 XM_011527655.1 XM_011527656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5232609 c.887_888insA p.D296fs*3 Chr.19: 18899718-18899719 on Build GRCh38
6494595 c.921C>A p.S307R Chr.19: 18899685-18899685 on Build GRCh38
7329159 c.897G>T p.R299S Chr.19: 18899709-18899709 on Build GRCh38
3692538 c.927A>C p.*309C Chr.19: 18899679-18899679 on Build GRCh38
4075770 c.912C>T p.Y304Y Chr.19: 18899694-18899694 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1009446552 Chr.19: 18899536-18899536 on Build GRCh38
rs769379786 Chr.19: 18899714-18899714 on Build GRCh38
rs149854108 Chr.19: 18899685-18899685 on Build GRCh38
rs770474643 Chr.19: 18899704-18899704 on Build GRCh38
rs776393167 Chr.19: 18899705-18899705 on Build GRCh38
rs1044823 Chr.19: 18899585-18899585 on Build GRCh38
rs966804879 Chr.19: 18899588-18899588 on Build GRCh38
rs949124667 Chr.19: 18899583-18899583 on Build GRCh38
rs377486646 Chr.19: 18899694-18899694 on Build GRCh38
rs751959133 Chr.19: 18899658-18899658 on Build GRCh38
rs764495624 Chr.19: 18899654-18899654 on Build GRCh38
rs777361082 Chr.19: 18899700-18899700 on Build GRCh38
rs534794917 Chr.19: 18899552-18899552 on Build GRCh38
rs376801368 Chr.19: 18899637-18899637 on Build GRCh38
rs565918834 Chr.19: 18899530-18899530 on Build GRCh38
rs757418856 Chr.19: 18899666-18899666 on Build GRCh38
rs916349648 Chr.19: 18899556-18899556 on Build GRCh38
rs1053663056 Chr.19: 18899620-18899620 on Build GRCh38
rs758937805 Chr.19: 18899708-18899708 on Build GRCh38
rs775154069 Chr.19: 18899715-18899715 on Build GRCh38
rs1022234172 Chr.19: 18899573-18899573 on Build GRCh38
rs781536955 Chr.19: 18899668-18899668 on Build GRCh38
rs745916838 Chr.19: 18899674-18899674 on Build GRCh38
rs753296377 Chr.19: 18899644-18899644 on Build GRCh38
rs934994233 Chr.19: 18899607-18899607 on Build GRCh38
rs746886291 Chr.19: 18899702-18899702 on Build GRCh38
rs775998127 Chr.19: 18899634-18899634 on Build GRCh38
rs765817112 Chr.19: 18899638-18899638 on Build GRCh38
rs1010890228 Chr.19: 18899559-18899559 on Build GRCh38
rs561587835 Chr.19: 18899569-18899569 on Build GRCh38
rs370888761 Chr.19: 18899653-18899653 on Build GRCh38
rs757311091 Chr.19: 18899694-18899694 on Build GRCh38
rs923568422 Chr.19: 18899589-18899589 on Build GRCh38
rs756229569 Chr.19: 18899677-18899677 on Build GRCh38
rs1046072339 Chr.19: 18899587-18899587 on Build GRCh38
rs973759610 Chr.19: 18899693-18899693 on Build GRCh38
rs772007066 Chr.19: 18899698-18899698 on Build GRCh38

View Full Product Details