Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14632842-14633113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE3
Entrez Gene ID
84658
Gene Name
adhesion G protein-coupled receptor E3
Gene Aliases
EMR3
UniGene
Hs.658712
Gene Chromosome Location
Chr.19: 14600175-14674918 on Build GRCh38
Transcripts
NM_001289158.1 NM_001289159.1 NM_032571.4 XM_011528374.2 XM_017027383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6465590 c.1602T>C p.L534L Chr.19: 14632962-14632962 on Build GRCh38
6465591 c.1601T>C p.L534P Chr.19: 14632963-14632963 on Build GRCh38
6208378 c.1643+1G>A p.? Chr.19: 14632920-14632920 on Build GRCh38
6017324 c.1643+9T>G p.? Chr.19: 14632912-14632912 on Build GRCh38
3282318 c.1607C>T p.S536F Chr.19: 14632957-14632957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2045912 Chr.19: 14633090-14633090 on Build GRCh38
rs763943954 Chr.19: 14633063-14633063 on Build GRCh38
rs142461083 Chr.19: 14632996-14632996 on Build GRCh38
rs762678750 Chr.19: 14633061-14633061 on Build GRCh38
rs997434813 Chr.19: 14632933-14632933 on Build GRCh38
rs770320312 Chr.19: 14633004-14633004 on Build GRCh38
rs543225515 Chr.19: 14633034-14633034 on Build GRCh38
rs1013692765 Chr.19: 14633059-14633059 on Build GRCh38
rs138384322 Chr.19: 14632979-14632979 on Build GRCh38
rs751604025 Chr.19: 14632990-14632990 on Build GRCh38
rs776318881 Chr.19: 14633030-14633030 on Build GRCh38
rs1022080015 Chr.19: 14632909-14632909 on Build GRCh38
rs752513062 Chr.19: 14633037-14633037 on Build GRCh38
rs947445050 Chr.19: 14632963-14632963 on Build GRCh38
rs760169835 Chr.19: 14632951-14632951 on Build GRCh38
rs776186924 Chr.19: 14632891-14632891 on Build GRCh38
rs781326971 Chr.19: 14632998-14632998 on Build GRCh38
rs749618477 Chr.19: 14633010-14633010 on Build GRCh38
rs560321303 Chr.19: 14632950-14632950 on Build GRCh38
rs921379037 Chr.19: 14632917-14632917 on Build GRCh38
rs763522270 Chr.19: 14632897-14632897 on Build GRCh38
rs147058520 Chr.19: 14633062-14633062 on Build GRCh38
rs772617742 Chr.19: 14632937-14632937 on Build GRCh38
rs975422775 Chr.19: 14632907-14632907 on Build GRCh38
rs769242670 Chr.19: 14632915-14632915 on Build GRCh38
rs968229331 Chr.19: 14632918-14632918 on Build GRCh38
rs554746691 Chr.19: 14633015-14633015 on Build GRCh38
rs377132894 Chr.19: 14633058-14633058 on Build GRCh38
rs982995988 Chr.19: 14632884-14632884 on Build GRCh38
rs770464933 Chr.19: 14633019-14633019 on Build GRCh38
rs773045534 Chr.19: 14633067-14633067 on Build GRCh38
rs533852689 Chr.19: 14632957-14632957 on Build GRCh38
rs769054109 Chr.19: 14633011-14633011 on Build GRCh38
rs764996847 Chr.19: 14632970-14632970 on Build GRCh38
rs751500719 Chr.19: 14633064-14633064 on Build GRCh38
rs553905155 Chr.19: 14632953-14632953 on Build GRCh38
rs764830342 Chr.19: 14633035-14633035 on Build GRCh38
rs759388525 Chr.19: 14632964-14632964 on Build GRCh38
rs373143666 Chr.19: 14632988-14632988 on Build GRCh38

View Full Product Details