Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87349000-87349273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROT
Entrez Gene ID
54677
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
COT
UniGene
Hs.125039
Gene Chromosome Location
Chr.7: 87345635-87399796 on Build GRCh38
Transcripts
NM_001143935.1 NM_001243745.1 NM_021151.3 XM_011516337.2 XM_017012370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6178521 c.104A>C p.Y35S Chr.7: 87349172-87349172 on Build GRCh38
1643397 c.96delA p.K34fs*6 Chr.7: 87349164-87349164 on Build GRCh38
4674769 c.95_96insA p.Y35fs*3 Chr.7: 87349163-87349164 on Build GRCh38
1452551 c.62C>A p.P21Q Chr.7: 87349130-87349130 on Build GRCh38
1202322 c.88G>T p.E30* Chr.7: 87349156-87349156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113592238 Chr.7: 87349158-87349158 on Build GRCh38
rs760951827 Chr.7: 87349081-87349081 on Build GRCh38
rs1044850408 Chr.7: 87349244-87349244 on Build GRCh38
rs11971666 Chr.7: 87349238-87349238 on Build GRCh38
rs770594340 Chr.7: 87349131-87349131 on Build GRCh38
rs140842977 Chr.7: 87349102-87349102 on Build GRCh38
rs147240633 Chr.7: 87349084-87349084 on Build GRCh38
rs756999481 Chr.7: 87349164-87349164 on Build GRCh38
rs752943012 Chr.7: 87349069-87349069 on Build GRCh38
rs779947503 Chr.7: 87349029-87349030 on Build GRCh38
rs777583546 Chr.7: 87349224-87349224 on Build GRCh38
rs925595133 Chr.7: 87349194-87349194 on Build GRCh38
rs754075882 Chr.7: 87349089-87349089 on Build GRCh38
rs775037264 Chr.7: 87349041-87349041 on Build GRCh38
rs757379228 Chr.7: 87349092-87349092 on Build GRCh38
rs201700761 Chr.7: 87349139-87349139 on Build GRCh38
rs778936531 Chr.7: 87349100-87349100 on Build GRCh38
rs754865059 Chr.7: 87349103-87349103 on Build GRCh38
rs769627937 Chr.7: 87349113-87349113 on Build GRCh38
rs777350070 Chr.7: 87349116-87349116 on Build GRCh38
rs748762185 Chr.7: 87349051-87349051 on Build GRCh38
rs369691197 Chr.7: 87349105-87349105 on Build GRCh38
rs761000290 Chr.7: 87349164-87349164 on Build GRCh38
rs765963910 Chr.7: 87349036-87349036 on Build GRCh38
rs575414855 Chr.7: 87349146-87349146 on Build GRCh38
rs776057015 Chr.7: 87349150-87349150 on Build GRCh38
rs778596623 Chr.7: 87349055-87349055 on Build GRCh38
rs201394030 Chr.7: 87349174-87349174 on Build GRCh38
rs374673999 Chr.7: 87349184-87349184 on Build GRCh38
rs143103247 Chr.7: 87349142-87349142 on Build GRCh38
rs368674447 Chr.7: 87349208-87349208 on Build GRCh38
rs765327773 Chr.7: 87349198-87349198 on Build GRCh38
rs375446390 Chr.7: 87349053-87349053 on Build GRCh38
rs748938256 Chr.7: 87349121-87349121 on Build GRCh38
rs1030180624 Chr.7: 87349119-87349119 on Build GRCh38
rs747831338 Chr.7: 87349107-87349107 on Build GRCh38
rs935493924 Chr.7: 87349196-87349196 on Build GRCh38
rs918086998 Chr.7: 87349226-87349226 on Build GRCh38
rs754050219 Chr.7: 87349176-87349176 on Build GRCh38
rs781060257 Chr.7: 87349206-87349206 on Build GRCh38
rs753873696 Chr.7: 87349029-87349032 on Build GRCh38
rs1052726930 Chr.7: 87349172-87349172 on Build GRCh38
rs802037 Chr.7: 87349031-87349031 on Build GRCh38
rs762439177 Chr.7: 87349033-87349033 on Build GRCh38
rs755900759 Chr.7: 87349216-87349216 on Build GRCh38
rs955405514 Chr.7: 87349135-87349135 on Build GRCh38
rs772636771 Chr.7: 87349149-87349149 on Build GRCh38
rs750445851 Chr.7: 87349203-87349203 on Build GRCh38
rs911185532 Chr.7: 87349148-87349148 on Build GRCh38
rs371392044 Chr.7: 87349144-87349144 on Build GRCh38

View Full Product Details