Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34489896-34490169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELF5
Entrez Gene ID
2001
Gene Name
E74 like ETS transcription factor 5
Gene Aliases
ESE2
UniGene
Hs.11713
Gene Chromosome Location
Chr.11: 34478791-34513800 on Build GRCh38
Transcripts
NM_001243080.1 NM_001243081.1 NM_001422.3 NM_198381.1 XM_017017308.1 XM_017017309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5017522 c.399delT p.F133fs*20 Chr.11: 34490046-34490046 on Build GRCh38
4032535 c.422C>T p.A141V Chr.11: 34490023-34490023 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377109616 Chr.11: 34489960-34489960 on Build GRCh38
rs768074835 Chr.11: 34490095-34490095 on Build GRCh38
rs895825654 Chr.11: 34490128-34490128 on Build GRCh38
rs779315222 Chr.11: 34490082-34490082 on Build GRCh38
rs755778429 Chr.11: 34489951-34489951 on Build GRCh38
rs145030473 Chr.11: 34490040-34490040 on Build GRCh38
rs565673341 Chr.11: 34490101-34490101 on Build GRCh38
rs750913133 Chr.11: 34490088-34490088 on Build GRCh38
rs960952991 Chr.11: 34489947-34489947 on Build GRCh38
rs552164354 Chr.11: 34490081-34490081 on Build GRCh38
rs754069457 Chr.11: 34490074-34490074 on Build GRCh38
rs2231825 Chr.11: 34489994-34489994 on Build GRCh38
rs756406296 Chr.11: 34489970-34489970 on Build GRCh38
rs1043800607 Chr.11: 34490113-34490113 on Build GRCh38
rs764328124 Chr.11: 34490067-34490067 on Build GRCh38
rs539338277 Chr.11: 34490121-34490121 on Build GRCh38
rs940775831 Chr.11: 34490043-34490043 on Build GRCh38
rs776181635 Chr.11: 34490034-34490034 on Build GRCh38
rs763052336 Chr.11: 34490052-34490052 on Build GRCh38
rs943895626 Chr.11: 34489990-34489990 on Build GRCh38
rs771476053 Chr.11: 34490001-34490001 on Build GRCh38
rs779404000 Chr.11: 34490094-34490094 on Build GRCh38
rs376040055 Chr.11: 34490093-34490093 on Build GRCh38
rs781109928 Chr.11: 34489989-34489989 on Build GRCh38
rs773189017 Chr.11: 34490110-34490110 on Build GRCh38
rs1014426419 Chr.11: 34490141-34490141 on Build GRCh38
rs532151103 Chr.11: 34490063-34490063 on Build GRCh38
rs999674754 Chr.11: 34489940-34489940 on Build GRCh38
rs368334489 Chr.11: 34490039-34490039 on Build GRCh38
rs777968155 Chr.11: 34489975-34489975 on Build GRCh38
rs746621726 Chr.11: 34490031-34490031 on Build GRCh38
rs760699423 Chr.11: 34490059-34490059 on Build GRCh38
rs755990303 Chr.11: 34490028-34490028 on Build GRCh38
rs773219474 Chr.11: 34490051-34490051 on Build GRCh38
rs910927304 Chr.11: 34489978-34489978 on Build GRCh38
rs754161410 Chr.11: 34489993-34489993 on Build GRCh38
rs371372901 Chr.11: 34490084-34490084 on Build GRCh38
rs375363670 Chr.11: 34490030-34490030 on Build GRCh38
rs767471452 Chr.11: 34490054-34490054 on Build GRCh38
rs769709171 Chr.11: 34490105-34490105 on Build GRCh38
rs199749742 Chr.11: 34490033-34490033 on Build GRCh38
rs1035600066 Chr.11: 34489946-34489946 on Build GRCh38
rs762406480 Chr.11: 34490055-34490055 on Build GRCh38
rs747809202 Chr.11: 34490103-34490103 on Build GRCh38
rs946333715 Chr.11: 34490108-34490108 on Build GRCh38
rs746376024 Chr.11: 34489995-34489995 on Build GRCh38

View Full Product Details