Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151717199-151717472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FATE1
Entrez Gene ID
89885
Gene Name
fetal and adult testis expressed 1
Gene Aliases
CT43, FATE
UniGene
Hs.301960
Gene Chromosome Location
Chr.X: 151716036-151723194 on Build GRCh38
Transcripts
NM_033085.2

SNP Detail

SNP ID Chromosome Location
rs765535480 Chr.X: 151717407-151717407 on Build GRCh38
rs189855448 Chr.X: 151717324-151717324 on Build GRCh38
rs764691725 Chr.X: 151717424-151717424 on Build GRCh38
rs766572542 Chr.X: 151717227-151717227 on Build GRCh38
rs138359911 Chr.X: 151717364-151717364 on Build GRCh38
rs147067761 Chr.X: 151717293-151717293 on Build GRCh38
rs752043405 Chr.X: 151717436-151717436 on Build GRCh38
rs758854056 Chr.X: 151717411-151717411 on Build GRCh38
rs753127743 Chr.X: 151717258-151717258 on Build GRCh38
rs768778324 Chr.X: 151717224-151717224 on Build GRCh38
rs774327171 Chr.X: 151717361-151717361 on Build GRCh38
rs373362524 Chr.X: 151717362-151717362 on Build GRCh38
rs776057205 Chr.X: 151717259-151717261 on Build GRCh38
rs767549871 Chr.X: 151717356-151717356 on Build GRCh38
rs372161224 Chr.X: 151717372-151717372 on Build GRCh38
rs755231513 Chr.X: 151717241-151717241 on Build GRCh38
rs754124594 Chr.X: 151717237-151717237 on Build GRCh38
rs773194679 Chr.X: 151717337-151717337 on Build GRCh38
rs780857408 Chr.X: 151717302-151717302 on Build GRCh38
rs769513189 Chr.X: 151717312-151717312 on Build GRCh38
rs758801183 Chr.X: 151717267-151717267 on Build GRCh38
rs202137219 Chr.X: 151717299-151717299 on Build GRCh38
rs778394151 Chr.X: 151717278-151717278 on Build GRCh38
rs757850277 Chr.X: 151717437-151717437 on Build GRCh38
rs768452944 Chr.X: 151717349-151717349 on Build GRCh38
rs774464435 Chr.X: 151717287-151717287 on Build GRCh38
rs759629114 Chr.X: 151717386-151717386 on Build GRCh38
rs752981867 Chr.X: 151717409-151717409 on Build GRCh38
rs765462123 Chr.X: 151717257-151717257 on Build GRCh38
rs566490903 Chr.X: 151717309-151717309 on Build GRCh38

View Full Product Details