Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 20038280-20038527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRNKL1
Entrez Gene ID
51340
Gene Name
crooked neck pre-mRNA splicing factor 1
Gene Aliases
CLF, CRN, Clf1, HCRN, MSTP021, SYF3
UniGene
Hs.171342
Gene Chromosome Location
Chr.20: 20034361-20056046 on Build GRCh38
Transcripts
NM_001278625.1 NM_001278626.1 NM_001278627.1 NM_001278628.1 NM_016652.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1024972 c.2117C>T p.T706M Chr.20: 20038362-20038362 on Build GRCh38
4996000 c.2098C>T p.R700W Chr.20: 20038381-20038381 on Build GRCh38
1716136 c.2113C>T p.R705W Chr.20: 20038366-20038366 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1032498283 Chr.20: 20038495-20038495 on Build GRCh38
rs780965619 Chr.20: 20038393-20038393 on Build GRCh38
rs530550457 Chr.20: 20038501-20038501 on Build GRCh38
rs746147865 Chr.20: 20038431-20038432 on Build GRCh38
rs545337876 Chr.20: 20038483-20038483 on Build GRCh38
rs369861927 Chr.20: 20038381-20038381 on Build GRCh38
rs775650884 Chr.20: 20038312-20038312 on Build GRCh38
rs764654665 Chr.20: 20038361-20038361 on Build GRCh38
rs371482244 Chr.20: 20038435-20038435 on Build GRCh38
rs199819737 Chr.20: 20038418-20038418 on Build GRCh38
rs753308068 Chr.20: 20038426-20038426 on Build GRCh38
rs200745538 Chr.20: 20038392-20038392 on Build GRCh38
rs915744156 Chr.20: 20038330-20038330 on Build GRCh38
rs7271629 Chr.20: 20038503-20038503 on Build GRCh38
rs897719363 Chr.20: 20038410-20038410 on Build GRCh38
rs76980563 Chr.20: 20038375-20038375 on Build GRCh38
rs968472565 Chr.20: 20038450-20038450 on Build GRCh38
rs574039596 Chr.20: 20038324-20038324 on Build GRCh38
rs749450469 Chr.20: 20038439-20038439 on Build GRCh38
rs778890988 Chr.20: 20038478-20038478 on Build GRCh38
rs563558951 Chr.20: 20038490-20038490 on Build GRCh38
rs757439711 Chr.20: 20038466-20038466 on Build GRCh38
rs762215398 Chr.20: 20038378-20038378 on Build GRCh38
rs754933661 Chr.20: 20038433-20038433 on Build GRCh38
rs377678635 Chr.20: 20038377-20038377 on Build GRCh38
rs763555385 Chr.20: 20038325-20038325 on Build GRCh38

View Full Product Details