Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48192625-48192872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMPD2
Entrez Gene ID
143162
Gene Name
FERM and PDZ domain containing 2
Gene Aliases
PDZD5C, PDZK4, PDZK5C
UniGene
Hs.664786
Gene Chromosome Location
Chr.10: 48156559-48274898 on Build GRCh38
Transcripts
NM_001018071.3 NM_001318191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5370681 c.2066_2067CC>AT p.S689Y Chr.10: 48192782-48192783 on Build GRCh38
4014363 c.2006G>A p.R669Q Chr.10: 48192843-48192843 on Build GRCh38
6667072 c.2020A>G p.I674V Chr.10: 48192829-48192829 on Build GRCh38
214105 c.2135A>T p.K712M Chr.10: 48192714-48192714 on Build GRCh38
5903749 c.2053G>A p.E685K Chr.10: 48192796-48192796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369560793 Chr.10: 48192657-48192657 on Build GRCh38
rs369496785 Chr.10: 48192728-48192728 on Build GRCh38
rs773597347 Chr.10: 48192830-48192830 on Build GRCh38
rs1039829789 Chr.10: 48192778-48192778 on Build GRCh38
rs767590164 Chr.10: 48192765-48192765 on Build GRCh38
rs374320487 Chr.10: 48192844-48192844 on Build GRCh38
rs115963581 Chr.10: 48192722-48192722 on Build GRCh38
rs116034788 Chr.10: 48192687-48192687 on Build GRCh38
rs201066967 Chr.10: 48192710-48192710 on Build GRCh38
rs762262160 Chr.10: 48192647-48192647 on Build GRCh38
rs747826265 Chr.10: 48192809-48192809 on Build GRCh38
rs778753992 Chr.10: 48192768-48192768 on Build GRCh38
rs760390374 Chr.10: 48192853-48192853 on Build GRCh38
rs776377372 Chr.10: 48192720-48192720 on Build GRCh38
rs755068594 Chr.10: 48192759-48192759 on Build GRCh38
rs74878516 Chr.10: 48192703-48192703 on Build GRCh38
rs146233973 Chr.10: 48192688-48192688 on Build GRCh38
rs758418695 Chr.10: 48192662-48192662 on Build GRCh38
rs758390268 Chr.10: 48192796-48192796 on Build GRCh38
rs763354849 Chr.10: 48192831-48192831 on Build GRCh38
rs768286419 Chr.10: 48192801-48192801 on Build GRCh38
rs757038094 Chr.10: 48192675-48192682 on Build GRCh38
rs973751233 Chr.10: 48192715-48192715 on Build GRCh38
rs1028240633 Chr.10: 48192789-48192789 on Build GRCh38
rs751613132 Chr.10: 48192754-48192754 on Build GRCh38
rs752968203 Chr.10: 48192766-48192766 on Build GRCh38
rs140347747 Chr.10: 48192727-48192727 on Build GRCh38
rs4838398 Chr.10: 48192752-48192752 on Build GRCh38
rs866413521 Chr.10: 48192771-48192771 on Build GRCh38
rs115774076 Chr.10: 48192691-48192691 on Build GRCh38
rs994888673 Chr.10: 48192780-48192780 on Build GRCh38
rs372691254 Chr.10: 48192705-48192705 on Build GRCh38
rs957021063 Chr.10: 48192685-48192685 on Build GRCh38
rs769403306 Chr.10: 48192825-48192825 on Build GRCh38
rs535329989 Chr.10: 48192787-48192787 on Build GRCh38
rs912294812 Chr.10: 48192739-48192739 on Build GRCh38
rs780097743 Chr.10: 48192666-48192666 on Build GRCh38
rs373611937 Chr.10: 48192661-48192661 on Build GRCh38
rs111320090 Chr.10: 48192797-48192797 on Build GRCh38
rs776093995 Chr.10: 48192806-48192806 on Build GRCh38
rs766909761 Chr.10: 48192753-48192753 on Build GRCh38
rs778648914 Chr.10: 48192692-48192692 on Build GRCh38
rs116457606 Chr.10: 48192686-48192686 on Build GRCh38
rs375396407 Chr.10: 48192668-48192668 on Build GRCh38
rs762017871 Chr.10: 48192721-48192721 on Build GRCh38
rs752777542 Chr.10: 48192846-48192846 on Build GRCh38
rs756943773 Chr.10: 48192767-48192767 on Build GRCh38
rs370193041 Chr.10: 48192843-48192843 on Build GRCh38
rs766819950 Chr.10: 48192832-48192832 on Build GRCh38
rs138806670 Chr.10: 48192702-48192702 on Build GRCh38
rs745721953 Chr.10: 48192795-48192795 on Build GRCh38
rs774866262 Chr.10: 48192840-48192840 on Build GRCh38

View Full Product Details