Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132453386-132453657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC13
Entrez Gene ID
23317
Gene Name
DnaJ heat shock protein family (Hsp40) member C13
Gene Aliases
PARK21, RME8
UniGene
Hs.12707
Gene Chromosome Location
Chr.3: 132417527-132539032 on Build GRCh38
Transcripts
NM_015268.3 XM_005247245.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6647229 c.722delA p.I243fs*13 Chr.3: 132453482-132453482 on Build GRCh38
4630829 c.665G>A p.R222H Chr.3: 132453425-132453425 on Build GRCh38
228366 c.751G>C p.V251L Chr.3: 132453605-132453605 on Build GRCh38
6647248 c.801G>A p.P267P Chr.3: 132453655-132453655 on Build GRCh38
5995200 c.779C>T p.T260I Chr.3: 132453633-132453633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773709434 Chr.3: 132453562-132453562 on Build GRCh38
rs367584768 Chr.3: 132453451-132453451 on Build GRCh38
rs202069439 Chr.3: 132453424-132453424 on Build GRCh38
rs754276059 Chr.3: 132453589-132453589 on Build GRCh38
rs754222838 Chr.3: 132453458-132453458 on Build GRCh38
rs964574934 Chr.3: 132453573-132453573 on Build GRCh38
rs762534279 Chr.3: 132453434-132453434 on Build GRCh38
rs543905953 Chr.3: 132453603-132453603 on Build GRCh38
rs148186584 Chr.3: 132453452-132453452 on Build GRCh38
rs573676097 Chr.3: 132453591-132453591 on Build GRCh38
rs777960726 Chr.3: 132453619-132453619 on Build GRCh38
rs747009692 Chr.3: 132453509-132453509 on Build GRCh38
rs976428756 Chr.3: 132453478-132453478 on Build GRCh38
rs79808031 Chr.3: 132453583-132453583 on Build GRCh38
rs569280440 Chr.3: 132453414-132453414 on Build GRCh38
rs774934666 Chr.3: 132453426-132453426 on Build GRCh38
rs769650097 Chr.3: 132453531-132453531 on Build GRCh38
rs757375779 Chr.3: 132453634-132453634 on Build GRCh38
rs765507211 Chr.3: 132453599-132453599 on Build GRCh38
rs1019996151 Chr.3: 132453527-132453527 on Build GRCh38
rs368364734 Chr.3: 132453582-132453582 on Build GRCh38
rs759315286 Chr.3: 132453418-132453418 on Build GRCh38
rs779112560 Chr.3: 132453480-132453480 on Build GRCh38
rs751003080 Chr.3: 132453438-132453438 on Build GRCh38
rs761262838 Chr.3: 132453576-132453576 on Build GRCh38
rs755295498 Chr.3: 132453464-132453464 on Build GRCh38
rs748261120 Chr.3: 132453494-132453494 on Build GRCh38
rs1001340878 Chr.3: 132453441-132453441 on Build GRCh38
rs777712064 Chr.3: 132453504-132453504 on Build GRCh38
rs965542250 Chr.3: 132453475-132453475 on Build GRCh38
rs907033081 Chr.3: 132453425-132453425 on Build GRCh38
rs776664452 Chr.3: 132453521-132453521 on Build GRCh38
rs139307487 Chr.3: 132453588-132453589 on Build GRCh38
rs558102985 Chr.3: 132453543-132453543 on Build GRCh38
rs7613098 Chr.3: 132453551-132453551 on Build GRCh38
rs1045647159 Chr.3: 132453421-132453421 on Build GRCh38
rs770974149 Chr.3: 132453513-132453513 on Build GRCh38
rs539705635 Chr.3: 132453525-132453525 on Build GRCh38
rs763495079 Chr.3: 132453437-132453437 on Build GRCh38
rs751616278 Chr.3: 132453628-132453628 on Build GRCh38
rs758519786 Chr.3: 132453609-132453609 on Build GRCh38
rs540403662 Chr.3: 132453556-132453556 on Build GRCh38
rs920073492 Chr.3: 132453617-132453617 on Build GRCh38

View Full Product Details