Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37021451-37021676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUPT20H
Entrez Gene ID
55578
Gene Name
SPT20 homolog, SAGA complex component
Gene Aliases
C13, C13orf19, FAM48A, FP757, P38IP, SPT20
UniGene
Hs.435815
Gene Chromosome Location
Chr.13: 37009312-37059714 on Build GRCh38
Transcripts
NM_001014286.2 NM_001278480.1 NM_001278481.1 NM_001278482.1 NM_017569.3 XM_005266447.2 XM_005266449.3 XM_005266451.3 XM_005266454.3 XM_005266455.2 XM_005266456.3 XM_005266457.3 XM_005266458.3 XM_005266460.2 XM_005266461.3 XM_005266462.4 XM_005266464.3 XM_005266465.2 XM_005266467.3 XM_017020653.1 XM_017020654.1 XM_017020655.1 XM_017020656.1 XM_017020657.1 XM_017020658.1 XM_017020659.1 XM_017020660.1 XM_017020661.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6483772 c.1665G>T p.G555G Chr.13: 37021599-37021599 on Build GRCh38
6580631 c.1801C>T p.Q601* Chr.13: 37021466-37021466 on Build GRCh38
696644 c.1664G>A p.G555E Chr.13: 37021600-37021600 on Build GRCh38
6139296 c.1753C>G p.L585V Chr.13: 37021511-37021511 on Build GRCh38
6521006 c.1664G>T p.G555V Chr.13: 37021600-37021600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755314663 Chr.13: 37021503-37021503 on Build GRCh38
rs779204624 Chr.13: 37021504-37021504 on Build GRCh38
rs766121706 Chr.13: 37021505-37021505 on Build GRCh38
rs780738673 Chr.13: 37021557-37021557 on Build GRCh38
rs1046838310 Chr.13: 37021501-37021501 on Build GRCh38
rs199796421 Chr.13: 37021517-37021517 on Build GRCh38
rs952919870 Chr.13: 37021482-37021482 on Build GRCh38
rs762614387 Chr.13: 37021610-37021610 on Build GRCh38
rs768534321 Chr.13: 37021630-37021630 on Build GRCh38
rs758816969 Chr.13: 37021518-37021518 on Build GRCh38
rs760465730 Chr.13: 37021485-37021485 on Build GRCh38
rs775230282 Chr.13: 37021594-37021594 on Build GRCh38
rs778105167 Chr.13: 37021524-37021524 on Build GRCh38
rs554703387 Chr.13: 37021638-37021638 on Build GRCh38
rs144899348 Chr.13: 37021567-37021567 on Build GRCh38
rs777339871 Chr.13: 37021646-37021647 on Build GRCh38
rs770406704 Chr.13: 37021553-37021553 on Build GRCh38
rs929696737 Chr.13: 37021499-37021499 on Build GRCh38
rs760803976 Chr.13: 37021480-37021480 on Build GRCh38
rs142518257 Chr.13: 37021537-37021537 on Build GRCh38
rs111384116 Chr.13: 37021473-37021473 on Build GRCh38
rs188364760 Chr.13: 37021495-37021495 on Build GRCh38
rs965419464 Chr.13: 37021509-37021509 on Build GRCh38
rs769272648 Chr.13: 37021574-37021574 on Build GRCh38
rs9603129 Chr.13: 37021565-37021565 on Build GRCh38
rs1018361579 Chr.13: 37021506-37021506 on Build GRCh38
rs761896125 Chr.13: 37021650-37021650 on Build GRCh38

View Full Product Details