Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23766120-23766345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHCHD10
Entrez Gene ID
400916
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
Gene Aliases
C22orf16, FTDALS2, IMMD, N27C7-4, SMAJ
UniGene
Hs.66915
Gene Chromosome Location
Chr.22: 23765834-23767972 on Build GRCh38
Transcripts
NM_001301339.1 NM_213720.2 NR_125755.1 NR_125756.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3552556 c.360C>T p.S120S Chr.22: 23766177-23766177 on Build GRCh38
4611904 c.283delC p.Q95fs*35 Chr.22: 23766254-23766254 on Build GRCh38
726171 c.406C>A p.H136N Chr.22: 23766131-23766131 on Build GRCh38
3668588 c.303C>T p.P101P Chr.22: 23766234-23766234 on Build GRCh38
5573710 c.262-1G>A p.? Chr.22: 23766276-23766276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776781535 Chr.22: 23766235-23766235 on Build GRCh38
rs867521564 Chr.22: 23766278-23766278 on Build GRCh38
rs777049622 Chr.22: 23766153-23766153 on Build GRCh38
rs199579266 Chr.22: 23766159-23766159 on Build GRCh38
rs533095236 Chr.22: 23766282-23766282 on Build GRCh38
rs750056280 Chr.22: 23766280-23766280 on Build GRCh38
rs761383051 Chr.22: 23766267-23766267 on Build GRCh38
rs776074927 Chr.22: 23766257-23766257 on Build GRCh38
rs1014921249 Chr.22: 23766277-23766277 on Build GRCh38
rs750293377 Chr.22: 23766205-23766205 on Build GRCh38
rs767301039 Chr.22: 23766269-23766269 on Build GRCh38
rs111677724 Chr.22: 23766251-23766251 on Build GRCh38
rs780072582 Chr.22: 23766220-23766220 on Build GRCh38
rs931085449 Chr.22: 23766188-23766188 on Build GRCh38
rs527737360 Chr.22: 23766231-23766231 on Build GRCh38
rs889489701 Chr.22: 23766214-23766214 on Build GRCh38
rs562212051 Chr.22: 23766230-23766230 on Build GRCh38
rs1021130344 Chr.22: 23766279-23766279 on Build GRCh38
rs377740688 Chr.22: 23766180-23766180 on Build GRCh38
rs564469294 Chr.22: 23766274-23766274 on Build GRCh38
rs9153 Chr.22: 23766225-23766225 on Build GRCh38
rs187396358 Chr.22: 23766177-23766177 on Build GRCh38
rs780959799 Chr.22: 23766183-23766183 on Build GRCh38
rs372902325 Chr.22: 23766165-23766165 on Build GRCh38
rs773772416 Chr.22: 23766264-23766264 on Build GRCh38
rs138183274 Chr.22: 23766207-23766207 on Build GRCh38
rs902121413 Chr.22: 23766268-23766268 on Build GRCh38
rs1000576672 Chr.22: 23766147-23766147 on Build GRCh38
rs374353973 Chr.22: 23766263-23766263 on Build GRCh38
rs763453989 Chr.22: 23766173-23766173 on Build GRCh38
rs760754380 Chr.22: 23766145-23766145 on Build GRCh38
rs762839189 Chr.22: 23766253-23766268 on Build GRCh38
rs765753241 Chr.22: 23766158-23766158 on Build GRCh38
rs371437007 Chr.22: 23766155-23766155 on Build GRCh38
rs771132146 Chr.22: 23766149-23766149 on Build GRCh38
rs759922090 Chr.22: 23766242-23766242 on Build GRCh38
rs773491269 Chr.22: 23766143-23766143 on Build GRCh38
rs964013488 Chr.22: 23766203-23766203 on Build GRCh38

View Full Product Details