Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51345700-51345973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA1
Entrez Gene ID
1990
Gene Name
chymotrypsin like elastase family member 1
Gene Aliases
ELA1
UniGene
Hs.348395
Gene Chromosome Location
Chr.12: 51328443-51346679 on Build GRCh38
Transcripts
NM_001971.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1705590 c.40G>A p.E14K Chr.12: 51345854-51345854 on Build GRCh38
3764428 c.30G>A p.Q10Q Chr.12: 51345864-51345864 on Build GRCh38
6369406 c.30G>T p.Q10H Chr.12: 51345864-51345864 on Build GRCh38
3753313 c.30G>C p.Q10H Chr.12: 51345864-51345864 on Build GRCh38
6312605 c.17-5C>A p.? Chr.12: 51345882-51345882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916133624 Chr.12: 51345934-51345934 on Build GRCh38
rs17860287 Chr.12: 51345864-51345864 on Build GRCh38
rs148155083 Chr.12: 51345737-51345737 on Build GRCh38
rs141103193 Chr.12: 51345829-51345829 on Build GRCh38
rs76499560 Chr.12: 51345948-51345948 on Build GRCh38
rs377124562 Chr.12: 51345796-51345796 on Build GRCh38
rs901160618 Chr.12: 51345807-51345807 on Build GRCh38
rs770677930 Chr.12: 51345884-51345884 on Build GRCh38
rs756313200 Chr.12: 51345835-51345835 on Build GRCh38
rs937660936 Chr.12: 51345898-51345898 on Build GRCh38
rs372331027 Chr.12: 51345776-51345776 on Build GRCh38
rs773945340 Chr.12: 51345889-51345891 on Build GRCh38
rs564811658 Chr.12: 51345939-51345939 on Build GRCh38
rs181750918 Chr.12: 51345834-51345834 on Build GRCh38
rs534392117 Chr.12: 51345856-51345856 on Build GRCh38
rs201804451 Chr.12: 51345855-51345855 on Build GRCh38
rs776129085 Chr.12: 51345931-51345931 on Build GRCh38
rs974710740 Chr.12: 51345949-51345949 on Build GRCh38
rs571468808 Chr.12: 51345840-51345840 on Build GRCh38
rs1024697706 Chr.12: 51345842-51345842 on Build GRCh38
rs74336876 Chr.12: 51345841-51345841 on Build GRCh38
rs772445683 Chr.12: 51345765-51345765 on Build GRCh38
rs763881505 Chr.12: 51345832-51345832 on Build GRCh38
rs759266978 Chr.12: 51345916-51345917 on Build GRCh38
rs903274002 Chr.12: 51345941-51345941 on Build GRCh38
rs531404251 Chr.12: 51345738-51345738 on Build GRCh38
rs956224671 Chr.12: 51345872-51345872 on Build GRCh38
rs747447671 Chr.12: 51345883-51345883 on Build GRCh38
rs866788056 Chr.12: 51345817-51345817 on Build GRCh38
rs866503837 Chr.12: 51345795-51345795 on Build GRCh38
rs765064498 Chr.12: 51345822-51345822 on Build GRCh38
rs999263570 Chr.12: 51345771-51345771 on Build GRCh38
rs745348426 Chr.12: 51345843-51345843 on Build GRCh38
rs749676963 Chr.12: 51345936-51345936 on Build GRCh38
rs200914206 Chr.12: 51345818-51345818 on Build GRCh38
rs199947566 Chr.12: 51345826-51345826 on Build GRCh38
rs551268679 Chr.12: 51345777-51345777 on Build GRCh38
rs981578201 Chr.12: 51345859-51345859 on Build GRCh38

View Full Product Details