Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43210196-43210469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB42
Entrez Gene ID
2038
Gene Name
erythrocyte membrane protein band 4.2
Gene Aliases
PA, SPH5
UniGene
Hs.368642
Gene Chromosome Location
Chr.15: 43197227-43225856 on Build GRCh38
Transcripts
NM_000119.2 NM_001114134.1 XM_005254225.1 XM_011521349.2 XM_011521350.2 XM_011521351.2 XM_011521352.2 XM_011521353.2 XM_011521354.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4054779 c.721G>A p.V241M Chr.15: 43210358-43210358 on Build GRCh38
6654791 c.642C>T p.F214F Chr.15: 43210437-43210437 on Build GRCh38
5054294 c.671G>A p.R224H Chr.15: 43210408-43210408 on Build GRCh38
244273 c.670C>T p.R224C Chr.15: 43210409-43210409 on Build GRCh38
1666254 c.688A>G p.K230E Chr.15: 43210391-43210391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181433780 Chr.15: 43210292-43210292 on Build GRCh38
rs768338326 Chr.15: 43210299-43210299 on Build GRCh38
rs764887771 Chr.15: 43210430-43210430 on Build GRCh38
rs373666732 Chr.15: 43210429-43210429 on Build GRCh38
rs142235741 Chr.15: 43210365-43210365 on Build GRCh38
rs760526823 Chr.15: 43210298-43210298 on Build GRCh38
rs199634098 Chr.15: 43210433-43210433 on Build GRCh38
rs764677935 Chr.15: 43210321-43210321 on Build GRCh38
rs199833430 Chr.15: 43210349-43210349 on Build GRCh38
rs769529879 Chr.15: 43210432-43210432 on Build GRCh38
rs535068940 Chr.15: 43210229-43210229 on Build GRCh38
rs761430208 Chr.15: 43210302-43210302 on Build GRCh38
rs139182437 Chr.15: 43210409-43210409 on Build GRCh38
rs115660852 Chr.15: 43210352-43210352 on Build GRCh38
rs967214409 Chr.15: 43210272-43210272 on Build GRCh38
rs758323919 Chr.15: 43210369-43210369 on Build GRCh38
rs776301800 Chr.15: 43210418-43210418 on Build GRCh38
rs561571428 Chr.15: 43210436-43210436 on Build GRCh38
rs958856930 Chr.15: 43210215-43210215 on Build GRCh38
rs752892136 Chr.15: 43210358-43210358 on Build GRCh38
rs768289851 Chr.15: 43210396-43210396 on Build GRCh38
rs1020030410 Chr.15: 43210263-43210263 on Build GRCh38
rs200274212 Chr.15: 43210317-43210317 on Build GRCh38
rs776157682 Chr.15: 43210313-43210313 on Build GRCh38
rs768899304 Chr.15: 43210437-43210437 on Build GRCh38
rs756167313 Chr.15: 43210359-43210359 on Build GRCh38
rs555481085 Chr.15: 43210244-43210244 on Build GRCh38
rs780732862 Chr.15: 43210232-43210232 on Build GRCh38
rs747737312 Chr.15: 43210424-43210424 on Build GRCh38
rs146348439 Chr.15: 43210408-43210408 on Build GRCh38
rs776355483 Chr.15: 43210321-43210321 on Build GRCh38
rs776240376 Chr.15: 43210298-43210298 on Build GRCh38
rs372856442 Chr.15: 43210296-43210296 on Build GRCh38
rs1034059684 Chr.15: 43210228-43210228 on Build GRCh38
rs1039614446 Chr.15: 43210301-43210301 on Build GRCh38
rs569639313 Chr.15: 43210217-43210217 on Build GRCh38
rs369798244 Chr.15: 43210329-43210329 on Build GRCh38
rs370874819 Chr.15: 43210351-43210351 on Build GRCh38
rs201923886 Chr.15: 43210366-43210366 on Build GRCh38
rs565635727 Chr.15: 43210260-43210260 on Build GRCh38
rs1011362812 Chr.15: 43210261-43210261 on Build GRCh38
rs72721553 Chr.15: 43210328-43210328 on Build GRCh38
rs750003231 Chr.15: 43210322-43210322 on Build GRCh38

View Full Product Details