Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 129908118-129908391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHSY3
Entrez Gene ID
337876
Gene Name
chondroitin sulfate synthase 3
Gene Aliases
CHSY2, CSS3
UniGene
Hs.213137
Gene Chromosome Location
Chr.5: 129903415-130186637 on Build GRCh38
Transcripts
NM_175856.4 XM_005271982.3 XM_005271983.3 XM_011543363.2 XM_011543365.2 XM_017009435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2150829 c.889C>G p.L297V Chr.5: 129908163-129908163 on Build GRCh38
1201187 c.1067A>G p.Q356R Chr.5: 129908341-129908341 on Build GRCh38
1736291 c.931G>T p.G311* Chr.5: 129908205-129908205 on Build GRCh38
6029884 c.902G>T p.G301V Chr.5: 129908176-129908176 on Build GRCh38
6888651 c.859G>A p.G287R Chr.5: 129908133-129908133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769259374 Chr.5: 129908275-129908275 on Build GRCh38
rs189874834 Chr.5: 129908320-129908320 on Build GRCh38
rs375577622 Chr.5: 129908184-129908184 on Build GRCh38
rs201625556 Chr.5: 129908274-129908274 on Build GRCh38
rs759731239 Chr.5: 129908307-129908307 on Build GRCh38
rs769383909 Chr.5: 129908202-129908202 on Build GRCh38
rs780697735 Chr.5: 129908272-129908272 on Build GRCh38
rs752938410 Chr.5: 129908259-129908259 on Build GRCh38
rs138111280 Chr.5: 129908153-129908153 on Build GRCh38
rs748841233 Chr.5: 129908282-129908282 on Build GRCh38
rs777729488 Chr.5: 129908322-129908322 on Build GRCh38
rs765623093 Chr.5: 129908256-129908256 on Build GRCh38
rs762201656 Chr.5: 129908222-129908222 on Build GRCh38
rs763048701 Chr.5: 129908319-129908319 on Build GRCh38
rs757078868 Chr.5: 129908266-129908266 on Build GRCh38
rs149892119 Chr.5: 129908230-129908230 on Build GRCh38
rs764502168 Chr.5: 129908253-129908253 on Build GRCh38
rs374883939 Chr.5: 129908280-129908280 on Build GRCh38
rs777273565 Chr.5: 129908306-129908306 on Build GRCh38
rs773380386 Chr.5: 129908286-129908286 on Build GRCh38
rs752597420 Chr.5: 129908150-129908150 on Build GRCh38
rs777808269 Chr.5: 129908156-129908156 on Build GRCh38
rs371776468 Chr.5: 129908263-129908263 on Build GRCh38
rs758264316 Chr.5: 129908260-129908260 on Build GRCh38
rs767478296 Chr.5: 129908349-129908349 on Build GRCh38
rs759675289 Chr.5: 129908243-129908243 on Build GRCh38
rs757065986 Chr.5: 129908342-129908342 on Build GRCh38
rs866599665 Chr.5: 129908238-129908239 on Build GRCh38
rs751549729 Chr.5: 129908264-129908264 on Build GRCh38
rs761045221 Chr.5: 129908231-129908231 on Build GRCh38
rs778864560 Chr.5: 129908144-129908144 on Build GRCh38
rs761257284 Chr.5: 129908287-129908287 on Build GRCh38
rs745528081 Chr.5: 129908201-129908201 on Build GRCh38
rs373792516 Chr.5: 129908318-129908318 on Build GRCh38
rs564978476 Chr.5: 129908325-129908325 on Build GRCh38
rs753910685 Chr.5: 129908235-129908235 on Build GRCh38
rs141140678 Chr.5: 129908315-129908315 on Build GRCh38
rs771398732 Chr.5: 129908288-129908288 on Build GRCh38
rs746901442 Chr.5: 129908162-129908162 on Build GRCh38
rs368190226 Chr.5: 129908217-129908217 on Build GRCh38
rs758224259 Chr.5: 129908285-129908285 on Build GRCh38
rs772299728 Chr.5: 129908229-129908229 on Build GRCh38
rs770499602 Chr.5: 129908168-129908168 on Build GRCh38

View Full Product Details