Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64932073-64932298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1
Entrez Gene ID
91612
Gene Name
churchill domain containing 1
Gene Aliases
C14orf52, My015, chch
UniGene
Hs.731537
Gene Chromosome Location
Chr.14: 64914361-64935368 on Build GRCh38
Transcripts
NM_001204063.1 NM_001204064.1 NM_145165.3 NM_001202558.1 NM_001202559.1

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001204063.1 NM_001204064.1 NM_145165.3 NM_001202558.1 NM_001202559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4921108 c.340T>C p.*114Q Chr.14: 64932228-64932228 on Build GRCh38
4577968 c.341T>G p.M114R Chr.14: 64932148-64932148 on Build GRCh38
4933110 c.301A>G p.I101V Chr.14: 64932189-64932189 on Build GRCh38
1241013 c.277G>A p.G93S Chr.14: 64932165-64932165 on Build GRCh38
6629152 c.285C>T p.A95A Chr.14: 64932173-64932173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763993146 Chr.14: 64932207-64932207 on Build GRCh38
rs773598743 Chr.14: 64932253-64932253 on Build GRCh38
rs200258159 Chr.14: 64932174-64932174 on Build GRCh38
rs1041299449 Chr.14: 64932146-64932146 on Build GRCh38
rs758090776 Chr.14: 64932226-64932226 on Build GRCh38
rs866552614 Chr.14: 64932166-64932166 on Build GRCh38
rs751863366 Chr.14: 64932185-64932185 on Build GRCh38
rs373972025 Chr.14: 64932187-64932187 on Build GRCh38
rs756213916 Chr.14: 64932203-64932203 on Build GRCh38
rs757003179 Chr.14: 64932212-64932212 on Build GRCh38
rs1004769320 Chr.14: 64932252-64932252 on Build GRCh38
rs768156423 Chr.14: 64932249-64932249 on Build GRCh38
rs760150920 Chr.14: 64932164-64932164 on Build GRCh38
rs45485893 Chr.14: 64932165-64932165 on Build GRCh38
rs746498285 Chr.14: 64932202-64932203 on Build GRCh38
rs778440646 Chr.14: 64932148-64932148 on Build GRCh38
rs554471925 Chr.14: 64932144-64932144 on Build GRCh38
rs774904587 Chr.14: 64932275-64932275 on Build GRCh38
rs749732606 Chr.14: 64932162-64932162 on Build GRCh38
rs753748838 Chr.14: 64932208-64932208 on Build GRCh38
rs762181692 Chr.14: 64932217-64932218 on Build GRCh38
rs745644580 Chr.14: 64932220-64932220 on Build GRCh38
rs745454070 Chr.14: 64932149-64932149 on Build GRCh38
rs375638413 Chr.14: 64932206-64932206 on Build GRCh38
rs759947536 Chr.14: 64932276-64932276 on Build GRCh38
rs776154153 Chr.14: 64932214-64932215 on Build GRCh38
rs749102184 Chr.14: 64932143-64932143 on Build GRCh38
rs911384213 Chr.14: 64932136-64932138 on Build GRCh38
rs370164458 Chr.14: 64932219-64932219 on Build GRCh38
rs748297128 Chr.14: 64932183-64932183 on Build GRCh38
rs768197977 Chr.14: 64932203-64932203 on Build GRCh38
rs777732976 Chr.14: 64932141-64932141 on Build GRCh38
rs771222663 Chr.14: 64932263-64932263 on Build GRCh38
rs79156210 Chr.14: 64932107-64932107 on Build GRCh38
rs756168630 Chr.14: 64932136-64932136 on Build GRCh38
rs373910233 Chr.14: 64932241-64932241 on Build GRCh38
rs1024617094 Chr.14: 64932205-64932205 on Build GRCh38
rs752703548 Chr.14: 64932122-64932122 on Build GRCh38
rs138406795 Chr.14: 64932231-64932231 on Build GRCh38
rs781380927 Chr.14: 64932118-64932118 on Build GRCh38
rs772308491 Chr.14: 64932186-64932186 on Build GRCh38
rs771598175 Chr.14: 64932150-64932150 on Build GRCh38
rs375937108 Chr.14: 64932258-64932258 on Build GRCh38
rs778953181 Chr.14: 64932173-64932173 on Build GRCh38
rs906202270 Chr.14: 64932245-64932245 on Build GRCh38

View Full Product Details