Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 154422441-154422702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNKSR3
Entrez Gene ID
154043
Gene Name
CNKSR family member 3
Gene Aliases
MAGI1
UniGene
Hs.16064
Gene Chromosome Location
Chr.6: 154405299-154510619 on Build GRCh38
Transcripts
NM_173515.2 XM_011535485.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6888672 c.840G>A p.E280E Chr.6: 154422611-154422611 on Build GRCh38
1672932 c.902C>T p.P301L Chr.6: 154422549-154422549 on Build GRCh38
5763638 c.923G>A p.R308Q Chr.6: 154422528-154422528 on Build GRCh38
483594 c.807G>A p.W269* Chr.6: 154422644-154422644 on Build GRCh38
74188 c.887C>A p.S296Y Chr.6: 154422564-154422564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756943868 Chr.6: 154422528-154422528 on Build GRCh38
rs117352540 Chr.6: 154422612-154422612 on Build GRCh38
rs748246477 Chr.6: 154422668-154422668 on Build GRCh38
rs772606447 Chr.6: 154422672-154422672 on Build GRCh38
rs929318088 Chr.6: 154422542-154422542 on Build GRCh38
rs768816305 Chr.6: 154422553-154422553 on Build GRCh38
rs142284063 Chr.6: 154422597-154422597 on Build GRCh38
rs368107025 Chr.6: 154422618-154422618 on Build GRCh38
rs751384658 Chr.6: 154422600-154422600 on Build GRCh38
rs781042760 Chr.6: 154422530-154422530 on Build GRCh38
rs780209855 Chr.6: 154422627-154422627 on Build GRCh38
rs773892810 Chr.6: 154422673-154422673 on Build GRCh38
rs777528000 Chr.6: 154422520-154422520 on Build GRCh38
rs138998489 Chr.6: 154422541-154422541 on Build GRCh38
rs540856434 Chr.6: 154422621-154422621 on Build GRCh38
rs375235956 Chr.6: 154422536-154422536 on Build GRCh38
rs754958727 Chr.6: 154422638-154422638 on Build GRCh38
rs752229556 Chr.6: 154422512-154422512 on Build GRCh38
rs757961840 Chr.6: 154422517-154422517 on Build GRCh38
rs774676860 Chr.6: 154422568-154422568 on Build GRCh38
rs1026453658 Chr.6: 154422566-154422566 on Build GRCh38
rs772261268 Chr.6: 154422573-154422573 on Build GRCh38
rs564517517 Chr.6: 154422577-154422577 on Build GRCh38
rs767502631 Chr.6: 154422602-154422602 on Build GRCh38
rs757213208 Chr.6: 154422601-154422601 on Build GRCh38
rs1005822871 Chr.6: 154422586-154422586 on Build GRCh38
rs770055394 Chr.6: 154422550-154422550 on Build GRCh38
rs756317238 Chr.6: 154422540-154422540 on Build GRCh38
rs763674094 Chr.6: 154422599-154422599 on Build GRCh38
rs766809340 Chr.6: 154422510-154422510 on Build GRCh38
rs751537312 Chr.6: 154422527-154422527 on Build GRCh38
rs943443463 Chr.6: 154422487-154422487 on Build GRCh38
rs369573689 Chr.6: 154422657-154422657 on Build GRCh38
rs749551092 Chr.6: 154422635-154422635 on Build GRCh38
rs868256496 Chr.6: 154422519-154422519 on Build GRCh38
rs748705440 Chr.6: 154422569-154422569 on Build GRCh38
rs920553668 Chr.6: 154422532-154422532 on Build GRCh38
rs773696288 Chr.6: 154422574-154422574 on Build GRCh38
rs555014730 Chr.6: 154422587-154422587 on Build GRCh38
rs775309729 Chr.6: 154422595-154422595 on Build GRCh38
rs906810425 Chr.6: 154422596-154422596 on Build GRCh38

View Full Product Details