Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232528294-232528565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRND
Entrez Gene ID
1144
Gene Name
cholinergic receptor nicotinic delta subunit
Gene Aliases
ACHRD, CMS2A, CMS3A, CMS3B, CMS3C, FCCMS, SCCMS
UniGene
Hs.156289
Gene Chromosome Location
Chr.2: 232526160-232537907 on Build GRCh38
Transcripts
NM_000751.2 NM_001256657.1 NM_001311195.1 NM_001311196.1 XM_011510524.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628798 c.362G>A p.G121D Chr.2: 232528509-232528509 on Build GRCh38
6356536 c.408C>T p.Y136Y Chr.2: 232528555-232528555 on Build GRCh38
1017829 c.386G>A p.C129Y Chr.2: 232528533-232528533 on Build GRCh38
1017828 c.305G>A p.R102H Chr.2: 232528323-232528323 on Build GRCh38
4164428 c.318C>G p.D106E Chr.2: 232528336-232528336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756306085 Chr.2: 232528451-232528451 on Build GRCh38
rs756324982 Chr.2: 232528522-232528522 on Build GRCh38
rs749350891 Chr.2: 232528457-232528457 on Build GRCh38
rs562896574 Chr.2: 232528456-232528456 on Build GRCh38
rs777541486 Chr.2: 232528419-232528419 on Build GRCh38
rs1045929925 Chr.2: 232528442-232528442 on Build GRCh38
rs185337948 Chr.2: 232528420-232528420 on Build GRCh38
rs769160054 Chr.2: 232528332-232528332 on Build GRCh38
rs752699014 Chr.2: 232528508-232528508 on Build GRCh38
rs765810417 Chr.2: 232528385-232528385 on Build GRCh38
rs760395222 Chr.2: 232528358-232528358 on Build GRCh38
rs370534958 Chr.2: 232528413-232528413 on Build GRCh38
rs147919651 Chr.2: 232528323-232528323 on Build GRCh38
rs755141289 Chr.2: 232528538-232528538 on Build GRCh38
rs772628400 Chr.2: 232528471-232528471 on Build GRCh38
rs770567115 Chr.2: 232528486-232528486 on Build GRCh38
rs773054021 Chr.2: 232528350-232528350 on Build GRCh38
rs776034693 Chr.2: 232528489-232528489 on Build GRCh38
rs765179580 Chr.2: 232528408-232528408 on Build GRCh38
rs575887548 Chr.2: 232528412-232528412 on Build GRCh38
rs764641517 Chr.2: 232528507-232528507 on Build GRCh38
rs2853451 Chr.2: 232528468-232528468 on Build GRCh38
rs772540361 Chr.2: 232528339-232528339 on Build GRCh38
rs55700159 Chr.2: 232528537-232528537 on Build GRCh38
rs779520432 Chr.2: 232528466-232528466 on Build GRCh38
rs753330133 Chr.2: 232528396-232528396 on Build GRCh38
rs775578238 Chr.2: 232528322-232528322 on Build GRCh38
rs751310035 Chr.2: 232528516-232528516 on Build GRCh38
rs759137435 Chr.2: 232528501-232528501 on Build GRCh38
rs763895244 Chr.2: 232528511-232528511 on Build GRCh38
rs760415811 Chr.2: 232528484-232528484 on Build GRCh38
rs773641076 Chr.2: 232528474-232528474 on Build GRCh38
rs748649626 Chr.2: 232528470-232528470 on Build GRCh38
rs780366075 Chr.2: 232528525-232528525 on Build GRCh38
rs759412532 Chr.2: 232528405-232528405 on Build GRCh38
rs764573389 Chr.2: 232528407-232528407 on Build GRCh38
rs971974036 Chr.2: 232528375-232528375 on Build GRCh38
rs770730077 Chr.2: 232528333-232528333 on Build GRCh38

View Full Product Details