Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119701497-119701770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIT
Entrez Gene ID
11113
Gene Name
citron rho-interacting serine/threonine kinase
Gene Aliases
CRIK, STK21
UniGene
Hs.119594
Gene Chromosome Location
Chr.12: 119685790-119877318 on Build GRCh38
Transcripts
NM_001206999.1 NM_007174.2 XM_006719206.2 XM_011537783.1 XM_011537784.1 XM_011537785.1 XM_011537787.1 XM_011537788.1 XM_011537789.1 XM_011537790.2 XM_011537791.2 XM_011537792.2 XM_017018735.1 XM_017018736.1 XM_017018737.1 XM_017018738.1 XM_017018739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1586187 c.5414-1G>T p.? Chr.12: 119701753-119701753 on Build GRCh38
3811186 c.5478C>T p.F1826F Chr.12: 119701688-119701688 on Build GRCh38
3456913 c.5418C>T p.F1806F Chr.12: 119701748-119701748 on Build GRCh38
6290045 c.5490C>T p.I1830I Chr.12: 119701676-119701676 on Build GRCh38
6290046 c.5364C>T p.I1788I Chr.12: 119701676-119701676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769593950 Chr.12: 119701687-119701687 on Build GRCh38
rs773246397 Chr.12: 119701688-119701688 on Build GRCh38
rs1018826933 Chr.12: 119701629-119701629 on Build GRCh38
rs752318456 Chr.12: 119701625-119701625 on Build GRCh38
rs766223378 Chr.12: 119701695-119701695 on Build GRCh38
rs761584754 Chr.12: 119701606-119701606 on Build GRCh38
rs201551867 Chr.12: 119701650-119701650 on Build GRCh38
rs763623625 Chr.12: 119701731-119701731 on Build GRCh38
rs763011355 Chr.12: 119701689-119701689 on Build GRCh38
rs746198257 Chr.12: 119701656-119701656 on Build GRCh38
rs765381193 Chr.12: 119701713-119701713 on Build GRCh38
rs987781674 Chr.12: 119701595-119701595 on Build GRCh38
rs776782740 Chr.12: 119701605-119701605 on Build GRCh38
rs751211586 Chr.12: 119701613-119701613 on Build GRCh38
rs574700924 Chr.12: 119701705-119701705 on Build GRCh38
rs561006897 Chr.12: 119701704-119701704 on Build GRCh38
rs138161610 Chr.12: 119701628-119701628 on Build GRCh38
rs776729880 Chr.12: 119701616-119701616 on Build GRCh38
rs968249730 Chr.12: 119701655-119701655 on Build GRCh38
rs372881627 Chr.12: 119701600-119701600 on Build GRCh38
rs920309812 Chr.12: 119701612-119701612 on Build GRCh38
rs765283743 Chr.12: 119701608-119701608 on Build GRCh38
rs545201342 Chr.12: 119701593-119701593 on Build GRCh38
rs779143986 Chr.12: 119701576-119701576 on Build GRCh38
rs559099333 Chr.12: 119701526-119701526 on Build GRCh38
rs947930495 Chr.12: 119701720-119701720 on Build GRCh38
rs777508949 Chr.12: 119701730-119701730 on Build GRCh38
rs768569538 Chr.12: 119701604-119701604 on Build GRCh38
rs149158991 Chr.12: 119701660-119701660 on Build GRCh38
rs543047397 Chr.12: 119701726-119701726 on Build GRCh38
rs567298155 Chr.12: 119701661-119701661 on Build GRCh38
rs1014720152 Chr.12: 119701570-119701570 on Build GRCh38
rs369065687 Chr.12: 119701617-119701617 on Build GRCh38
rs368957852 Chr.12: 119701603-119701603 on Build GRCh38
rs748315119 Chr.12: 119701678-119701678 on Build GRCh38
rs768924501 Chr.12: 119701675-119701675 on Build GRCh38
rs758358701 Chr.12: 119701706-119701706 on Build GRCh38
rs746291860 Chr.12: 119701588-119701588 on Build GRCh38
rs776565133 Chr.12: 119701676-119701676 on Build GRCh38
rs1020741303 Chr.12: 119701567-119701567 on Build GRCh38

View Full Product Details