Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42586716-42586985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM134C
Entrez Gene ID
162427
Gene Name
family with sequence similarity 134 member C
Gene Aliases
-
UniGene
Hs.632262
Gene Chromosome Location
Chr.17: 42579508-42609427 on Build GRCh38
Transcripts
NM_178126.3 NR_026697.1 XM_011524440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658482 c.504+2T>C p.? Chr.17: 42586763-42586763 on Build GRCh38
5813885 c.440T>A p.V147D Chr.17: 42586829-42586829 on Build GRCh38
7418757 c.427C>A p.H143N Chr.17: 42586842-42586842 on Build GRCh38
6658485 c.389T>C p.V130A Chr.17: 42586880-42586880 on Build GRCh38
1683269 c.472_473insGT p.V158fs*16 Chr.17: 42586796-42586797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756942284 Chr.17: 42586903-42586903 on Build GRCh38
rs754319953 Chr.17: 42586739-42586739 on Build GRCh38
rs556993035 Chr.17: 42586843-42586843 on Build GRCh38
rs758775508 Chr.17: 42586872-42586872 on Build GRCh38
rs754002348 Chr.17: 42586861-42586861 on Build GRCh38
rs780684428 Chr.17: 42586912-42586912 on Build GRCh38
rs552720553 Chr.17: 42586873-42586873 on Build GRCh38
rs746608852 Chr.17: 42586796-42586796 on Build GRCh38
rs574406491 Chr.17: 42586894-42586894 on Build GRCh38
rs755553255 Chr.17: 42586768-42586768 on Build GRCh38
rs150766247 Chr.17: 42586897-42586897 on Build GRCh38
rs202004998 Chr.17: 42586820-42586820 on Build GRCh38
rs776997508 Chr.17: 42586895-42586895 on Build GRCh38
rs570204374 Chr.17: 42586777-42586777 on Build GRCh38
rs760928569 Chr.17: 42586857-42586857 on Build GRCh38
rs775276368 Chr.17: 42586839-42586839 on Build GRCh38
rs1024942822 Chr.17: 42586874-42586874 on Build GRCh38
rs189259624 Chr.17: 42586774-42586774 on Build GRCh38
rs758760931 Chr.17: 42586775-42586775 on Build GRCh38
rs769182655 Chr.17: 42586929-42586929 on Build GRCh38
rs995722405 Chr.17: 42586814-42586814 on Build GRCh38
rs745441621 Chr.17: 42586923-42586923 on Build GRCh38
rs769817006 Chr.17: 42586830-42586830 on Build GRCh38
rs200843863 Chr.17: 42586840-42586840 on Build GRCh38
rs779270721 Chr.17: 42586769-42586769 on Build GRCh38
rs148877628 Chr.17: 42586860-42586860 on Build GRCh38
rs753080144 Chr.17: 42586871-42586871 on Build GRCh38
rs1032352671 Chr.17: 42586937-42586938 on Build GRCh38
rs375573712 Chr.17: 42586939-42586939 on Build GRCh38
rs780713729 Chr.17: 42586815-42586815 on Build GRCh38
rs1039381489 Chr.17: 42586808-42586808 on Build GRCh38
rs759525101 Chr.17: 42586862-42586862 on Build GRCh38
rs768410342 Chr.17: 42586911-42586911 on Build GRCh38
rs898124913 Chr.17: 42586810-42586810 on Build GRCh38
rs749175625 Chr.17: 42586941-42586941 on Build GRCh38
rs770451394 Chr.17: 42586798-42586798 on Build GRCh38
rs765842588 Chr.17: 42586863-42586863 on Build GRCh38
rs372724510 Chr.17: 42586855-42586855 on Build GRCh38
rs768495003 Chr.17: 42586842-42586842 on Build GRCh38

View Full Product Details