Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76333963-76334236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC158
Entrez Gene ID
339965
Gene Name
coiled-coil domain containing 158
Gene Aliases
-
UniGene
Hs.529680
Gene Chromosome Location
Chr.4: 76313039-76421920 on Build GRCh38
Transcripts
NM_001042784.1 XM_005262974.4 XM_006714204.3 XM_011531908.2 XM_011531909.2 XM_011531910.2 XM_011531911.1 XM_011531912.2 XM_011531913.1 XM_011531914.2 XM_011531915.2 XM_011531916.2 XM_011531917.1 XM_017008160.1 XM_017008161.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3428663 c.2779G>T p.E927* Chr.4: 76334053-76334053 on Build GRCh38
5642821 c.2724C>A p.L908L Chr.4: 76334108-76334108 on Build GRCh38
6621779 c.2713A>C p.K905Q Chr.4: 76334119-76334119 on Build GRCh38
5060473 c.2739C>T p.S913S Chr.4: 76334093-76334093 on Build GRCh38
3605970 c.2706G>A p.R902R Chr.4: 76334126-76334126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765722805 Chr.4: 76334053-76334053 on Build GRCh38
rs747853062 Chr.4: 76333991-76333991 on Build GRCh38
rs745774601 Chr.4: 76334023-76334023 on Build GRCh38
rs756745687 Chr.4: 76334068-76334068 on Build GRCh38
rs776544642 Chr.4: 76334051-76334051 on Build GRCh38
rs769445662 Chr.4: 76334021-76334021 on Build GRCh38
rs751171490 Chr.4: 76334067-76334067 on Build GRCh38
rs780134177 Chr.4: 76334077-76334077 on Build GRCh38
rs200087196 Chr.4: 76334025-76334025 on Build GRCh38
rs749080081 Chr.4: 76334031-76334031 on Build GRCh38
rs1043279678 Chr.4: 76334009-76334009 on Build GRCh38
rs781011177 Chr.4: 76334007-76334007 on Build GRCh38
rs750312135 Chr.4: 76334187-76334188 on Build GRCh38
rs771004429 Chr.4: 76334047-76334047 on Build GRCh38
rs866990920 Chr.4: 76334121-76334121 on Build GRCh38
rs745978201 Chr.4: 76334200-76334200 on Build GRCh38
rs754936497 Chr.4: 76334178-76334178 on Build GRCh38
rs200243245 Chr.4: 76333996-76333996 on Build GRCh38
rs113022439 Chr.4: 76334057-76334057 on Build GRCh38
rs752423971 Chr.4: 76334198-76334198 on Build GRCh38
rs114878412 Chr.4: 76334093-76334093 on Build GRCh38
rs767062503 Chr.4: 76334143-76334143 on Build GRCh38
rs775903125 Chr.4: 76334055-76334055 on Build GRCh38
rs763746027 Chr.4: 76334060-76334060 on Build GRCh38
rs778248483 Chr.4: 76334187-76334187 on Build GRCh38
rs191689378 Chr.4: 76334163-76334163 on Build GRCh38
rs775921599 Chr.4: 76334124-76334124 on Build GRCh38
rs374157248 Chr.4: 76334085-76334085 on Build GRCh38
rs780041629 Chr.4: 76334026-76334026 on Build GRCh38
rs769811878 Chr.4: 76334202-76334202 on Build GRCh38
rs747081719 Chr.4: 76334035-76334035 on Build GRCh38
rs781393638 Chr.4: 76334099-76334099 on Build GRCh38
rs370220929 Chr.4: 76334135-76334135 on Build GRCh38
rs760141058 Chr.4: 76334052-76334052 on Build GRCh38
rs756176859 Chr.4: 76334074-76334074 on Build GRCh38
rs376108896 Chr.4: 76334127-76334127 on Build GRCh38
rs781208195 Chr.4: 76334199-76334199 on Build GRCh38
rs574121898 Chr.4: 76334169-76334169 on Build GRCh38
rs768965221 Chr.4: 76334129-76334129 on Build GRCh38
rs745935143 Chr.4: 76334101-76334101 on Build GRCh38
rs761573901 Chr.4: 76334141-76334141 on Build GRCh38
rs750015585 Chr.4: 76334150-76334150 on Build GRCh38
rs754771967 Chr.4: 76334091-76334091 on Build GRCh38
rs772489421 Chr.4: 76334034-76334034 on Build GRCh38
rs370480373 Chr.4: 76334098-76334098 on Build GRCh38
rs897655866 Chr.4: 76334186-76334186 on Build GRCh38
rs756762692 Chr.4: 76334001-76334001 on Build GRCh38
rs756730042 Chr.4: 76333993-76333993 on Build GRCh38
rs778819264 Chr.4: 76334179-76334179 on Build GRCh38
rs778173692 Chr.4: 76334092-76334092 on Build GRCh38
rs937136612 Chr.4: 76334126-76334126 on Build GRCh38
rs745557636 Chr.4: 76334198-76334198 on Build GRCh38
rs749877494 Chr.4: 76334072-76334072 on Build GRCh38
rs373455626 Chr.4: 76334108-76334108 on Build GRCh38
rs748330853 Chr.4: 76334147-76334147 on Build GRCh38
rs753787829 Chr.4: 76334171-76334171 on Build GRCh38
rs767092600 Chr.4: 76334069-76334069 on Build GRCh38

View Full Product Details